Skip to Content
Merck
All Photos(1)

Key Documents

NIST2702

Inorganics in marine sediment

NIST® SRM® 2702

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 50 g

manufacturer/tradename

NIST®

application(s)

environmental

format

matrix material

General description

This Standard Reference Material (SRM) is marine sediment collected at the mouth of the Baltimore Harbor, Baltimore, MD and is intended for use in evaluating analytical methods for the determination of selected elements in marine or fresh water sediment and similar matrices. All of the constituents in SRM 2702, for which certified, reference, and information values are provided, were naturally present in the sediment material before processing. A unit of SRM 2702 consists of a bottle containing 50 g of radiation-sterilized, freeze-dried sediment material. For more information, please refer to the SDS and COA.

SRM 2702_cert

SRM 2702_SDS

Other Notes

Certified for the analytes listed below.
Aluminum (Al), Nickel (Ni), Antimony (Sb), Phosphorus (P), Arsenic (As), Potassium (K), Barium (Ba), Rubidium (Rb), Cadmium (Cd), Scandium (Sc), Cerium (Ce), Sodium (Na), Chromium (Cr), Strontium (Sr), Cobalt (Co), Thallium (Tl), Lanthanum (La), Thorium (Th), Lead (Pb), Titanium (Ti), Manganese (Mn), Vanadium (V), Mercury (Hg), Zinc (Zn)

Reference/Informational value is provided for the following analytes.
Calcium (Ca), Molybdenum (Mo), Copper (Cu), Selenium (Se), Gallium (Ga), Silver (Ag), Magnesium (Mg), Tin (Sn), Beryllium (Be), Neodymium (Nd), Carbon (C), Niobium (Nb), Samarium (Sm), Cesium (Cs), Sulfur (S), Hafnium (Hf), Tungsten (W), Iron (Fe), Uranium (U), Lithium (Li)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service