Skip to Content
Merck
All Photos(1)

Key Documents

NIST1546A

Meat homogenate

NIST® SRM® 1546a

Synonym(s):

Meat Sample Homogenate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 4 cans x 85 g

manufacturer/tradename

NIST®

format

matrix material

General description

Application


  • Quality Analysis and Biocontrol Activity: Research on the chemical composition and biocontrol activity of different essential oils, including meat homogenate, against soil-borne fungal pathogens demonstrates its efficacy in agricultural pest management and provides insights into sustainable farming practices (Akdeniz et al., 2024).

  • Antimicrobial Activity Studies: The essential oil of Mentha suaveolens, which includes components derived from meat homogenate, is analyzed for antimicrobial activity, showcasing its potential in pharmaceutical and cosmetic applications for enhancing product safety and efficacy (Afrokh et al., 2024).

  • Fish Freshness Assessment: Utilization of meat homogenate in developing a hypoxanthine enzyme sensor for assessing fish freshness. This application is critical for the seafood industry to ensure product quality and consumer safety (Wang et al., 2024).

  • Food Preservation Research: Meat homogenate is used in the study of garlic essential oil nanoemulsions for coating preservation of chilled fresh pork. This research is vital for extending the shelf life of meat products and improving food safety (Zhang et al., 2024).

  • Proteomic Analysis in Viral Infection: A study employs meat homogenate for label-free quantitative analysis of pig liver proteome post Hepatitis E virus infection, aiding in the understanding of viral impacts on livestock and contributing to the development of better veterinary health strategies (Martino et al., 2024).

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Ash, Calories, Carbohydrates, Fat / Saturated Fat, Phosphate, Protein, Solids
Amino acids: Alanine, Arginine, Aspartic acid, Cystine, Glutamic acid, Glycine, Histidine, Hydroxyproline, Isoleucine, Leucine, Lysine, Methionine, Phenylalanine, Proline, Serine, Taurine, Threonine, Tryptophan, Tyrosine, Valine
Lipids and Fatty acids: Capric acid, Lauric acid, Myristic acid, Myristoleic acid, Palmitic acid, Palmitoleic acid, Stearic acid, Oleic acid, Vaccenic acid, Linoleic acid, a-Linolenic acid, Arachidic acid, Gondoic acid, Arachidonic acid, Behenic acid, Lignoceric acid, Nervonic acid, Cholesterol, (Z,Z)-11,14-Eicosadienoic acid (C20:2 n-6), (Z,Z,Z)-11,14,17-Eicosatrienoic acid (C20:3 n-3), cis-Monounsaturated Fat, cis-Polyunsaturated Fat, Dihomo-γ-linolenic acid, DGLA, Elaidic acid, Erucic acid, Margaric acid, Margaroleic acid, Total cis-C18:1, Total cis-C18:2, Total cis-C20:1, Total cis-C22:4, Total cis-C22:5, Total trans Fat, Total trans-C18:1, Total trans-C18:2, Total trans-C18:2 conjugated, Total ϖ-3 Fatty acids, Total ϖ-6 Fatty acids, trans-Vaccenic acid, ϖ-Linolenic acid
Trace Elements: Barium (Ba), Boron (B), Calcium (Ca), Chorine, Copper (Cu), Iron (Fe), Magnesium (Mg), Manganese (Mn), Molybdenum (Mo), Potassium (K), Rubidium (Rb), Selenium, Sodium (Na), Strontium, Zinc (Zn)
Vitamins: 25-Hydroxyvitamin D3, Carnitine, Cholecalciferol (Vitamin D3), Choline, Niacin (Vitamin B3), Niacinamide (Vitamin B3), Pentadecanoic acid (C15:0), Pyridoxine (Vitamin B6), Riboflavin (Vitamin B2), Thiamine (Vitamin B1), Total Vitamin B12 by Microbiological Assay, Total Vitamin B3 as Niacinamide, Total Vitamin B5 by Microbiological Assay, Total Vitamin B6 as Pyridoxine, Total Vitamin B6 by Microbiological Assay

Matrix Group: Meat

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service