Skip to Content
Merck
All Photos(1)

Key Documents

N6142

Supelco

Nabumetone

analytical standard

Synonym(s):

4-(6-Methoxy-2-naphthyl)-2-butanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16O2
CAS Number:
Molecular Weight:
228.29
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

COc1ccc2cc(CCC(C)=O)ccc2c1

InChI

1S/C15H16O2/c1-11(16)3-4-12-5-6-14-10-15(17-2)8-7-13(14)9-12/h5-10H,3-4H2,1-2H3

InChI key

BLXXJMDCKKHMKV-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Nonselective non-steroidal anti-inflammatory drug (NSAID)

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Crothers et al.
International journal of pharmaceutics, 358(1-2), 303-306 (2008-04-18)
The solubilisation of two poorly soluble drugs, furosemide and nabumetone, in micellar solutions of diblock copolymers of ethylene oxide and styrene oxide has been studied at 25 and 37 degrees C and solubilisation capacities compared with published values for griseofulvin
Kaori Matsumoto et al.
Biological & pharmaceutical bulletin, 34(5), 734-739 (2011-05-03)
The cytochrome P450 (CYP) isoforms that catalyze the oxidation metabolism of 6-methoxy-2-napthylacetic acid (6-MNA), an active metabolite of nabumetone, were studied in rats and humans. Using an extractive reversed-phase HPLC assay with fluorescence detection, monophasic Michaelis-Menten kinetics was obtained for
Solubility of triamcinolone acetonide and other anti-inflammatory drugs in silicone oil: implications for Therapeutic Efficacy.
José Carlos Pastor et al.
Retina (Philadelphia, Pa.), 28(9), 1247-1250 (2008-07-17)
S Paul et al.
JNMA; journal of the Nepal Medical Association, 48(174), 121-125 (2010-04-15)
Aceclofenac, a NSAID is widely used in the treatment of pain and inflammation associated with osteoarthritis. Nabumetone, a recently developed preferential cyclo-oxygenase 2 inhibitor has also proved to be equally effective. The present study was undertaken to evaluate the 'real
Applicability of LC/MS/MS assay for 6-methoxy-2-napthylacetic acid to support the bioequivalence study of nabumetone-comments on the research work of Patel et al. (2008).
Nuggehally R Srinivas
Biomedical chromatography : BMC, 23(6), 674-675 (2009-03-12)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service