Skip to Content
Merck
All Photos(3)

Key Documents

N5514

Sigma-Aldrich

Nitro Blue Tetrazolium

tablet

Synonym(s):

Nitrotetrazolium Blue Chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

tablet

Quality Level

color

yellow

pH

4.5-5.5

solubility

soluble, clear, yellow

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[Cl-].[Cl-].COc1cc(ccc1-[n+]2nc(nn2-c3ccc(cc3)[N+]([O-])=O)-c4ccccc4)-c5ccc(c(OC)c5)-[n+]6nc(nn6-c7ccc(cc7)[N+]([O-])=O)-c8ccccc8

InChI

1S/C40H30N10O6.2ClH/c1-55-37-25-29(13-23-35(37)47-43-39(27-9-5-3-6-10-27)41-45(47)31-15-19-33(20-16-31)49(51)52)30-14-24-36(38(26-30)56-2)48-44-40(28-11-7-4-8-12-28)42-46(48)32-17-21-34(22-18-32)50(53)54;;/h3-26H,1-2H3;2*1H/q+2;;/p-2

InChI key

FSVCQIDHPKZJSO-UHFFFAOYSA-L

General description

Nitro Blue Tetrazolium is a tetrazole compound with two tetrazole rings.

Application

Nitro Blue Tetrazolium has been used:
  • to measure superoxide radicals
  • in lactate dehydrogenase quantification assay
  • for blot development in western blotting analysis
  • in NADPH-d histochemistry
  • as a reagent to analyse the properties of murine intestinal mucins by electrophoresis and histology
  • in immunohistochemistry

Quantity

Contains 10 mg substrate per tablet.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transient expression of a bovine leukemia virus envelope glycoprotein in plants by a recombinant TBSV vector
Zhumabek AT, et al.
Journal of Virological Methods, 255, 1-7 (2018)
Night light-adaptation strategies for photosynthetic apparatus in yellow-poplar (Liriodendron tulipifera L.) exposed to artificial night lighting
Kwak MJ, et al.
Forests, 9(2), 74-74 (2018)
Luttmann W
Immunology, The Experimenter Series (2006)
Paclitaxel inhibits expression of neuronal nitric oxide synthase and prevents mitochondrial dysfunction in spinal ventral horn in rats after C7 spinal root avulsion
Onler MDF
Turkish Neurosurgery, 25(4), 617-624 (2015)
Marine invertebrates cross phyla comparisons reveal highly conserved immune machinery.
Oren M
Immunology, 218, 484-484 (2013)

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service