Skip to Content
Merck
All Photos(1)

Key Documents

M7655

Supelco

Mesterolone

analytical standard

Synonym(s):

1α-Methylandrostan-17β-ol-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O2
CAS Number:
Molecular Weight:
304.47
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

[H][C@@]12CC[C@@]3([H])[C@]4([H])CC[C@H](O)[C@@]4(C)CC[C@]3([H])[C@@]1(C)[C@@H](C)CC(=O)C2

InChI

1S/C20H32O2/c1-12-10-14(21)11-13-4-5-15-16-6-7-18(22)19(16,2)9-8-17(15)20(12,13)3/h12-13,15-18,22H,4-11H2,1-3H3/t12-,13-,15-,16-,17-,18-,19-,20-/m0/s1

InChI key

UXYRZJKIQKRJCF-TZPFWLJSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 - Lact. - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D de Boer et al.
The Journal of steroid biochemistry and molecular biology, 42(3-4), 411-419 (1992-05-01)
Before including the detection of the methyl-5 alpha-dihydrotestosterones mesterolone (1 alpha-methyl-17 beta-hydroxy-5 alpha-androstan-3-one) and drostanolone (2 alpha-methyl-17 beta-hydroxy-5 alpha-androstan-3-one) in doping control procedures, their urinary metabolites were characterized by gas chromatography/mass spectrometry. Several metabolites were found after enzymatic hydrolysis and
M Iqbal Choudhary et al.
Natural product research, 22(15), 1289-1296 (2008-11-22)
Microbial transformation of two anabolic steroids, ethylestrenol (1) and nandrolone (2), were carried out. Ethylestrenol (1), when incubated with Rhizopus stolonifer (TSY 0471), yielded two oxidative metabolites named 17alpha-ethyl-3beta,17beta-dihydroxy-19-norndrost-4-ene (3) and 17alpha-ethyl-17beta-hydroxy-19-norandrost-4-en-3-one (4), while incubation of compound 2 with the
F Jockenhövel et al.
Metabolism: clinical and experimental, 48(5), 590-596 (1999-05-25)
We investigated whether the androgen type or application mode or testosterone (T) serum levels influence serum lipids and lipoprotein levels differentially in 55 hypogonadal men randomly assigned to the following treatment groups: mesterolone 100 mg orally daily ([MES] n =
Karina Fontana et al.
Cell and tissue research, 339(2), 411-420 (2009-11-11)
The effect of mesterolone and intensive treadmill training (6 weeks, 5 days/week, means: 15.82 m/min and 45.8 min/day) in Achilles tendon remodeling was evaluated. Sedentary mice treated with mesterolone (Sed-M) or vehicle (Sed-C, placebo/control) and corresponding exercised (Ex-M and Ex-C)
Karina Fontana et al.
Journal of electron microscopy, 59(2), 153-164 (2009-10-27)
Light and electron microscopy and quantitative morphometry were used to determine the effects of exercise and mesterolone on the soleus muscles of mice. Both exercise and mesterolone caused a significant hypertrophy of extrafusal muscle fibres. The hypertrophy of Type I

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service