Skip to Content
Merck
All Photos(1)

Key Documents

H0100000

Haloperidol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-[4-(4-Chlorophenyl)-4-hydroxy-1-piperidinyl]-1-(4-fluorophenyl)-1-butanone, 4-[4-(4-Chlorophenyl)-4-hydroxypiperidino]-4′-fluorobutyrophenone, 4-[4-(p-Chlorophenyl)-4-hydroxypiperidino]-4′-fluorobutyrophenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23ClFNO2
CAS Number:
Molecular Weight:
375.86
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

haloperidol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC1(CCN(CCCC(=O)c2ccc(F)cc2)CC1)c3ccc(Cl)cc3

InChI

1S/C21H23ClFNO2/c22-18-7-5-17(6-8-18)21(26)11-14-24(15-12-21)13-1-2-20(25)16-3-9-19(23)10-4-16/h3-10,26H,1-2,11-15H2

InChI key

LNEPOXFFQSENCJ-UHFFFAOYSA-N

Gene Information

human ... HTR2A(3356)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Haloperidol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dan Shan et al.
Schizophrenia research, 144(1-3), 1-8 (2013-01-30)
Glutamate transporters facilitate the buffering, clearance and cycling of glutamate and play an important role in maintaining synaptic and extrasynaptic glutamate levels. Alterations in glutamate transporter expression may lead to abnormal glutamate neurotransmission contributing to the pathophysiology of schizophrenia. In
Norma Adán et al.
The Journal of clinical investigation, 123(9), 3902-3913 (2013-08-03)
Chondrocytes are the only cells in cartilage, and their death by apoptosis contributes to cartilage loss in inflammatory joint diseases, such as rheumatoid arthritis (RA). A putative therapeutic intervention for RA is the inhibition of apoptosis-mediated cartilage degradation. The hormone
Mark C Fok et al.
International journal of geriatric psychiatry, 30(4), 333-344 (2015-02-03)
To summarize the effect of antipsychotics for preventing postoperative delirium. We conducted a literature search using Medline, EMBASE, Cochrane Central Register of Controlled Trials, Cochrane Database of Systematic Reviews and clinicaltrials.gov. We included randomized controlled trials of adults undergoing surgery
Gerard W K Hugenholtz et al.
The Journal of clinical psychiatry, 67(6), 897-903 (2006-07-20)
To determine the doses of haloperidol as a comparator drug in randomized controlled trials (RCTs) with atypical antipsychotics in patients with schizophrenia and to compare these doses with the officially recommended doses for haloperidol in the United States and the
Jan Booij et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(4), 647-649 (2014-03-08)
A recent (123)I-FP-CIT ((123)-I-N-ω-fluoropropyl-2β-carbomethoxy-3β-(4-iodophenyl)nortropane) SPECT study on rats suggested that a single 1 mg/kg dose of the antipsychotic haloperidol induces enough dopamine release to compete with (123)I-FP-CIT for binding to the dopamine transporter. Taking into account the far-reaching consequences of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service