Skip to Content
Merck
All Photos(1)

Key Documents

E0550000

Epirubicin hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4′-Epidoxorubicin hydrochloride, Epidoxorubicin hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29NO11 · HCl
CAS Number:
Molecular Weight:
579.98
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

epirubicin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.COc1cccc2C(=O)c3c(O)c4C[C@](O)(C[C@H](O[C@H]5C[C@H](N)[C@@H](O)[C@H](C)O5)c4c(O)c3C(=O)c12)C(=O)CO

InChI

1S/C27H29NO11.ClH/c1-10-22(31)13(28)6-17(38-10)39-15-8-27(36,16(30)9-29)7-12-19(15)26(35)21-20(24(12)33)23(32)11-4-3-5-14(37-2)18(11)25(21)34;/h3-5,10,13,15,17,22,29,31,33,35-36H,6-9,28H2,1-2H3;1H/t10-,13-,15-,17-,22-,27-;/m0./s1

InChI key

MWWSFMDVAYGXBV-FGBSZODSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Epirubicin hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Cell-permeable anthracycline antitumor antibiotic. Antineoplastic. A stereoisomer of doxorubicin that exhibits reduced cardiotoxicity. Its antitumor actions are mediated by targeting topoisomerase II.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Muta. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aman U Buzdar et al.
The Lancet. Oncology, 14(13), 1317-1325 (2013-11-19)
Neoadjuvant chemotherapy with trastuzumab for patients with HER2-positive breast cancer can produce a pathological complete response in the breast in 30-65% of patients. We investigated the effect of the timing of trastuzumab administration with anthracycline and taxane neoadjuvant chemotherapy. This
D Groheux et al.
British journal of cancer, 109(5), 1157-1164 (2013-08-15)
Pathologic complete response (pCR) to neoadjuvant treatment (NAT) is associated with improved survival of patients with HER2+ breast cancer. We investigated the ability of interim positron emission tomography (PET) regarding early prediction of pathology outcomes. During 61 months, consecutive patients
Chikako Suzuki et al.
Medical oncology (Northwood, London, England), 30(1), 415-415 (2013-01-17)
The aim of this was to determine whether the change of size observed at the first response evaluation after initiation of first-line combination chemotherapy correlates with overall survival (OS) in patients with metastatic breast cancer (MBC). The change in size
Sarah N Hamilton et al.
International journal of radiation oncology, biology, physics, 87(4), 719-725 (2013-09-05)
To review the treatment and outcomes of patients with primary cutaneous B-cell lymphoma (CBCL). Clinical characteristics, treatment, and outcomes were analyzed for all patients referred to our institution from 1981 through 2011 with primary CBCL without extracutaneous or distant nodal
Hung-Wei Yang et al.
Biomaterials, 34(29), 7204-7214 (2013-06-27)
Low accumulation of chemotherapeutic agent in tumor tissue and multidrug resistance (MDR) present a major obstacle to curing cancer treatment. Therefore, how to combine several therapeutics in one system is a key issue to overcome the problem. Here, we demonstrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service