Skip to Content
Merck
All Photos(1)

Key Documents

BP1075

Mesalazine

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

5-Aminosalicylic acid, 5-AS, 5-Amino-2-hydroxybenzoic acid, Mesalamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3-2-(OH)CO2H
CAS Number:
Molecular Weight:
153.14
Beilstein:
2090421
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

mesalazine, mesalamine

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

mp

275-280 °C (dec.) (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

Nc1ccc(O)c(c1)C(O)=O

InChI

1S/C7H7NO3/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,9H,8H2,(H,10,11)

InChI key

KBOPZPXVLCULAV-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mesalazine BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Mesalazine Suppositories
  • Mesalazine Foam Enema
  • Mesalazine Enema
  • Mesalazine Prolonged-release Tablets
  • Mesalazine Gastro-resistant Tablets
  • Mesalazine Prolonged-release Granules
  • 5-Aminosalicylic acid is a peroxidase substrate suitable for use in ELISA procedures. This substrate produces a soluble end product that is brown in color and can be read spectrophotometrically at 450 nm. The reaction may be stopped with 3 N NaOH and read at 550 nm.

    Packaging

    Unit quantity: 125 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    related product

    Product No.
    Description
    Pricing

    Pictograms

    Exclamation mark

    Signal Word

    Warning

    Hazard Statements

    Hazard Classifications

    Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

    Target Organs

    Respiratory system

    Storage Class Code

    11 - Combustible Solids

    WGK

    WGK 2

    Flash Point(F)

    536.0 °F

    Flash Point(C)

    280 °C


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Jingyi Hu et al.
    Journal of ethnopharmacology, 266, 113394-113394 (2020-09-18)
    Inflammatory bowel disease (IBD) is a chronic and relapsing inflammatory disease of the gastrointestinal tract, consisting of ulcerative colitis (UC) and Crohn's disease (CD). Gut microbiota and their metabolites may play a role in the pathogen of IBD, especially of

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service