Skip to Content
Merck
All Photos(1)

Key Documents

B5524

Supelco

Benzydamine hydrochloride

analytical standard

Synonym(s):

1-Benzyl-3-(3-[dimethylamino]propoxy)-1H-indazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23N3O · HCl
CAS Number:
Molecular Weight:
345.87
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

Cl.CN(C)CCCOc1nn(Cc2ccccc2)c3ccccc13

InChI

1S/C19H23N3O.ClH/c1-21(2)13-8-14-23-19-17-11-6-7-12-18(17)22(20-19)15-16-9-4-3-5-10-16;/h3-7,9-12H,8,13-15H2,1-2H3;1H

InChI key

HNNIWKQLJSNAEQ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Capolongo et al.
Journal of veterinary pharmacology and therapeutics, 33(4), 341-346 (2010-07-22)
Benzydamine (BZ), a weak base and an indazole derivative with analgesic and antipyretic properties used in human and veterinary medicine, is metabolized in human, rat, cattle and rabbit to a wide range of metabolites. One of the main metabolites, BZ
S Rossi et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 74(2), 248-254 (2009-10-27)
The aim of the present work was the development of a thermally sensitive mucoadhesive gel based on chitosan derivatives for the treatment of oral mucositis. Trimethyl chitosan (TMC) and methylpyrrolidinone chitosan (MPC) were considered. They were mixed with glycerophosphate (GP)
Yildiz Yilmaz et al.
Xenobiotica; the fate of foreign compounds in biological systems, 49(5), 503-512 (2018-04-26)
The expression of flavin-containing monooxygenase (FMO) varies extensively between human and commonly used preclinical species such as rat and mouse. The aim of this study was to investigate the pulmonary FMO activity in rat using benzydamine. Furthermore, the contribution of
Koray Soygun et al.
The journal of advanced prosthodontics, 9(3), 200-207 (2017-07-07)
In this study, three bioceramic materials, [IPS Empress CAD (Ivoclar), IPS e.max CAD (Ivoclar), and Lava Ultimate CAD (3M ESPE)] were treated with three commercial mouthrinses [Listerine, Tantum Verde, and Klorhex]; and changes in colour reflectance and surface roughness values
Ali Fuat Erciyas et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 48(10), 2577-2580 (2010-07-06)
In this study, genotoxicity of two mouthwash products (chlorexidin, benzidamine-HCl) were investigated in the Drosophila Wing-Spot Test which makes use of the wing cell markers multiple wing hairs (mwh) and flare (flr) and detects both mitotic recombination and various types

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service