Skip to Content
Merck
All Photos(1)

Key Documents

B018

Supelco

WB-4101 hydrochloride

analytical standard

Synonym(s):

2-[(2,6-Dimethoxyphenoxyethyl)aminomethyl]-1,4-benzodioxane hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23NO5 · HCl
CAS Number:
Molecular Weight:
381.85
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

Cl.COc1cccc(OC)c1OCCNCC2COc3ccccc3O2

InChI

1S/C19H23NO5.ClH/c1-21-17-8-5-9-18(22-2)19(17)23-11-10-20-12-14-13-24-15-6-3-4-7-16(15)25-14;/h3-9,14,20H,10-13H2,1-2H3;1H

InChI key

KAHMEWANVDFFCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Ichishima et al.
British journal of pharmacology, 161(1), 193-206 (2010-08-20)
Volume-regulated anion channels (VRACs) play an important role in cell-volume regulation. alpha(1)-Adrenoceptor stimulation by phenylephrine (PE) suppressed the hypotonic activation of VRAC current in mouse ventricular cells and regulatory volume decrease (RVD) was also absent in PE-treated cells. We examined
Jin O-Uchi et al.
Circulation research, 102(11), 1378-1388 (2008-05-10)
We examined the effect of alpha(1)-adrenoceptor subtype-specific stimulation on L-type Ca2+ current (I(Ca)) and elucidated the subtype-specific intracellular mechanisms for the regulation of L-type Ca2+ channels in isolated rat ventricular myocytes. We confirmed the protein expression of alpha(1A)- and alpha(1B)-adrenoceptor
W-X Yuan et al.
Neuroscience, 158(4), 1616-1624 (2008-12-11)
Activation of the descending noradrenergic system inhibits nociceptive transmission in the spinal cord. Although both alpha(1)- and alpha(2)-adrenoceptors in the spinal cord are involved in the modulation of nociceptive transmission, it is not clear how alpha(1)-adrenoceptors regulate excitatory and inhibitory
Josie R T Silva et al.
The journal of pain : official journal of the American Pain Society, 12(1), 51-60 (2010-06-18)
We evaluated the effectiveness of intrathecal antagonists of α1- (WB4101) and α2- (idazoxan) adrenoceptors and serotonergic (methysergide), opioid (naloxone), muscarinic (atropine), GABA(A) (bicuculline) and GABA(B) (phaclofen) receptors in blocking 2- or 100-Hz electroacupuncture (EA)-induced analgesia (EAIA) in the rat tail-flick
Laura Fumagalli et al.
European journal of medicinal chemistry, 58, 184-191 (2012-11-06)
Unichiral 8-substituted analogues of 2-[(2-(2,6-dimethoxyphenoxy)ethyl)aminomethyl]-1,4-benzodioxane (WB4101) were synthesized and tested for binding affinity at cloned human α(1a)-, α(1b)-and α(1d)-adrenoreceptor (α(1a)-, α(1b)-and α(1d)-AR) and at native rat 5-HT(1A) receptor and for antagonist affinity at α(1A)-, α(1B)-and α(1D)-AR and at α(2A/D)-AR. Among

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service