Skip to Content
Merck
All Photos(1)

Key Documents

74900

Supelco

1-Octene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH=CH2
CAS Number:
Molecular Weight:
112.21
Beilstein:
1734497
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.9 (vs air)

vapor pressure

36 mmHg ( 38 °C)

Assay

≥99.5% (GC)

autoignition temp.

446 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

3.9 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.408 (lit.)
n20/D 1.408
n20/D 1.409 (lit.)

bp

122-123 °C (lit.)

mp

−101 °C (lit.)

density

0.715 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care
petroleum

format

neat

SMILES string

CCCCCCC=C

InChI

1S/C8H16/c1-3-5-7-8-6-4-2/h3H,1,4-8H2,2H3

InChI key

KWKAKUADMBZCLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Octene is a linear α-olefin and an important comonomer in the production of linear low-density polyethylene (LLDPE).
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ethylene tetramerization: a new route to produce 1-octene in exceptionally high selectivities
Bollmann A, et al.
Journal of the American Chemical Society, 126(45), 14712-14713 (2004)
Emma B Hager et al.
Dalton transactions (Cambridge, England : 2003), 41(45), 13927-13935 (2012-10-02)
Water-soluble dendritic ligands based on tris-2-(5-sulfonato salicylaldimine ethyl)amine (5) and DAB-(5-sulfonato salicylaldimine) (6) (DAB = diaminobutane) were synthesized by means of Schiff base condensation and sulfonation reactions. These dendritic ligands were fully characterized by (1)H NMR, (13)C NMR and FT-IR
A cartridge system for organometallic catalysis: sequential catalysis and separation using supercritical carbon dioxide to switch phases.
Maurizio Solinas et al.
Angewandte Chemie (International ed. in English), 44(15), 2291-2295 (2005-03-12)
Cooperative ligand effects in phase-switching homogeneous catalysts.
Molise Mokhadinyana et al.
Angewandte Chemie (International ed. in English), 51(7), 1648-1652 (2012-01-13)
Aqueous-phase hydroformylation of 1-octene: styrene latices as phase-transfer agents.
Katharina Kunna et al.
Angewandte Chemie (International ed. in English), 45(43), 7289-7292 (2006-10-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service