Skip to Content
Merck
All Photos(1)

Key Documents

74582

Supelco

Chromium Standard for ICP

TraceCERT®, 1 g/L Cr in nitric acid (nominal concentration), prepared with Cr(NO3)3 x 9H2O

Synonym(s):

Chromium Calibration Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

composition

prepared with Cr(NO3)3 x 9H2O

concentration

1 g/L Cr in nitric acid (nominal concentration)

technique(s)

ICP: suitable

application(s)

industrial qc
pharmaceutical

format

single component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at https://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Application

Chromium ICP standard can be used as a calibration standard in the elemental analysis of contaminated water by inductively coupled plasma-atomic emissionspectrometry (ICP-AES).

Analysis Note

1 g/L Cr in 2% nitric acid, prepared with high purity Cr(NO3)3.9H2O, HNO3, and water

Other Notes

For a complete product listing of our TraceCERT® range of CRMs for ICP and AAS, technical information and example certificates please visit our ICP & AAS standards website

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microporous ceramic membrane technology for the removal of arsenic and chromium ions from contaminated water
Pagana A, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 110, 150-156 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service