Skip to Content
Merck
All Photos(4)

Key Documents

63675

Supelco

(+)-Menthone

greener alternative

analytical standard

Synonym(s):

(1S,4R)-p-Menthan-3-one, (2R,5S)-2-Isopropyl-5-methylcyclohexanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
2041367
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (sum of enantiomers, GC)

optical activity

[α]20/D +29±1°, neat

optical purity

enantiomeric ratio: ≥99:1 (GC)

shelf life

limited shelf life, expiry date on the label

greener alternative product score

old score: 43
new score: 15
Find out more about DOZN™ Scoring

greener alternative product characteristics

Atom Economy
Safer Solvents and Auxiliaries
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.450

bp

209-210 °C (lit.)

density

0.893 g/mL at 20 °C (lit.)

application(s)

food and beverages

format

neat

greener alternative category

storage temp.

2-8°C

SMILES string

CC(C)[C@H]1CC[C@H](C)CC1=O

InChI

1S/C10H18O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-9H,4-6H2,1-3H3/t8-,9+/m0/s1

InChI key

NFLGAXVYCFJBMK-DTWKUNHWSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Atom Economy”, “Safer Solvents and Auxiliaries” and "Enhanced Energy Efficiency". Click here to view its DOZN scorecard.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: mentha

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui Ma et al.
Plant physiology and biochemistry : PPB, 158, 363-371 (2020-11-28)
Cinnamomum camphora (L.) Presl, rich in terpenoids, is an important commercial plant. The monoterpenes borneol and camphor are highly desired compounds that have been widely and diversely used in medicine and spices since ancient times. However, the key enzymes in
Yi-Hsuan Lee et al.
The Canadian journal of infectious diseases & medical microbiology = Journal canadien des maladies infectieuses et de la microbiologie medicale, 2018, 8347403-8347403 (2018-06-01)
Essential oils from the dried spikes of Nepeta tenuifolia (Benth) are obtained by steam distillation. Pulegone was identified as the main component in the spikes of N. tenuifolia through analysis, with greater than 85% purity obtained in this study. The
Filitsa Karamaouna et al.
Journal of insect science (Online), 13, 142-142 (2014-04-29)
The vine mealybug, Planococcus ficus (Signoret) (Hemiptera: Pseudococcidae), is a pest in grape vine growing areas worldwide. The essential oils from the following aromatic plants were tested for their insecticidal activity against P. ficus: peppermint, Mentha piperita L. (Lamiales: Lamiaceae)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service