Skip to Content
Merck
All Photos(1)

Key Documents

63661

Supelco

(+)-Menthofuran

analytical standard

Synonym(s):

(6R)-4,5,6,7-Tetrahydro-3,6-dimethylbenzofuran, (R)-3,6-Dimethyl-4,5,6,7-tetrahydrobenzofuran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
Beilstein:
2729
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (sum of enantiomers, GC)

optical activity

[α]20/D +100±2°, neat

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.484

bp

204-206 °C (lit.)

density

0.97 g/mL at 20 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H]1CCc2c(C)coc2C1

InChI

1S/C10H14O/c1-7-3-4-9-8(2)6-11-10(9)5-7/h6-7H,3-5H2,1-2H3/t7-/m1/s1

InChI key

YGWKXXYGDYYFJU-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(+)-Menthofuran is a metabolite of pulegone, found in mint plants, which is basically produced from (+)–pulegone by menthofuran synthase enzyme, under certain conditions of stress like low sunlight, limited moisture and nutrient deficiencies.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: mentha

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Council of Europe
Chemically-defined Flavouring Substances (2000)
Rahman-U-A
Studies in Natural Products Chemistry, Volume 37, 37 (2012)
(R)-(+)-Menthofuran is a potent, mechanism-based inactivator of human liver cytochrome P450 2A6
Khojasteh - Bakht.CS
Drug Metabolism and Disposition, 26(7), 701-704 (1998)
Maria De Lucia et al.
The Journal of organic chemistry, 72(26), 10123-10129 (2007-11-30)
The reaction chemistry of menthofuran (1), a toxic furan terpenoid from various mint oils, with nitric acid and nitrous acid has been investigated. Treatment of 1 with nitric acid afforded a 1:1 mixture of the bisfuran derivatives 5 and 6
S Cyrus Khojasteh et al.
Chemical research in toxicology, 23(11), 1824-1832 (2010-10-16)
Menthofuran is a monoterpene present in mint plants that is oxidized by mammalian cytochrome P450 (CYP) to hepatotoxic metabolites. Evidence has been presented that p-cresol and other unusual oxidative products are metabolites of menthofuran in rats and that p-cresol may

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service