Skip to Content
Merck
All Photos(1)

Key Documents

44243

Supelco

Dodecyltrimethylammonium hydrogen sulfate

suitable for ion pair chromatography, LiChropur, ≥99.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11N(CH3)3HSO4
CAS Number:
Molecular Weight:
325.51
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NB.21

description

anionic

Quality Level

Assay

≥99.0% (T)

form

solid

quality

LiChropur

technique(s)

ion pair chromatography: suitable

λ

10 % in H2O

UV absorption

λ: 210 nm Amax: 0.1
λ: 240 nm Amax: 0.06
λ: 280 nm Amax: 0.04
λ: 310 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for filter test

SMILES string

OS([O-])(=O)=O.CCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C15H34N.H2O4S/c1-5-6-7-8-9-10-11-12-13-14-15-16(2,3)4;1-5(2,3)4/h5-15H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

KSZDKSNRUYOJHR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service