Skip to Content
Merck
All Photos(1)

Key Documents

43929

Supelco

Perfluorodecanoic acid

analytical standard

Synonym(s):

Nonadecafluorodecanoic acid, PFAS, Nonadecafluorocapric acid, Perfluorocapric acid, Perfluorodecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)8CO2H
CAS Number:
Molecular Weight:
514.08
Beilstein:
1810811
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

suitable for ASTM® 7968
suitable for ASTM® 7979
suitable for DIN 38407-42
suitable for EPA 1633
suitable for EPA 533
suitable for EPA 537.1
suitable for EPA 8327
suitable for EPA ACB B21-02
suitable for EPA ACB B23-05b
suitable for EPA OTM-45
suitable for ISO 21675 2019

reg. compliance

suitable for FDA C-010.02

vapor pressure

~10 mmHg ( 0 °C)

Assay

≥98.0% (GC)
98.0-102.0% (T)

shelf life

limited shelf life, expiry date on the label

bp

218 °C/740 mmHg (lit.)

mp

77-81 °C (lit.)

storage temp.

2-8°C

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C10HF19O2/c11-2(12,1(30)31)3(13,14)4(15,16)5(17,18)6(19,20)7(21,22)8(23,24)9(25,26)10(27,28)29/h(H,30,31)

InChI key

PCIUEQPBYFRTEM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Perfluorodecanoic acid is an environmental pollutant, belonging to the perfluorochemical (PFCs) class of compounds. These compounds are widely used as surfactants, polymer and plastic additives, wetting agents, corrosion inhibitors, lubricants, foam-fire extinguishants, etc.

Application

Perfluorodecanoic acid may be used as an analytical reference standard for the determination of the analyte in the following:       
  • biological samples using high-performance liquid chromatography (HPLC) with fluorescence detection     
  • whole blood samples using high-performance liquid chromatography-negative electrospray single quadrupole mass spectrometry (HPLC/ES-MS)        
  • environmental water samples using high-performance liquid chromatography-electrospray tandem mass spectrometry (HPLC/ESI-MS/MS)

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Lact. - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of perfluorinated compounds in environmental water samples by high-performance liquid chromatography-electrospray tandem mass spectrometry using surfactant-coated Fe3O4 magnetic nanoparticles as adsorbents
Zhao X, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 98(2), 207-214 (2011)
Determination of perfluorinated carboxylic acids in biological samples by high-performance liquid chromatography
Ohya T, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 720(1-2), 1-7 (1998)
Development of a solid-phase extraction-HPLC/single quadrupole MS method for quantification of perfluorochemicals in whole blood
Karrman A, et al.
Analytical Chemistry, 77(3), 864-870 (2005)

Articles

Liquid Chromatography–Mass Spectrometry (LC-MS) separation of 33 PFAS (Per- and poly-fluoroalkyl substances) using a superficially porous silica particles (SPP) based Ascentis® Express C18 column.

Liquid Chromatography–Mass Spectrometry (LC-MS) separation of 33 PFAS (Per- and poly-fluoroalkyl substances) using a superficially porous silica particles (SPP) based Ascentis® Express C18 column.

Liquid Chromatography–Mass Spectrometry (LC-MS) separation of 33 PFAS (Per- and poly-fluoroalkyl substances) using a superficially porous silica particles (SPP) based Ascentis® Express C18 column.

Liquid Chromatography–Mass Spectrometry (LC-MS) separation of 33 PFAS (Per- and poly-fluoroalkyl substances) using a superficially porous silica particles (SPP) based Ascentis® Express C18 column.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service