Skip to Content
Merck
All Photos(2)

Key Documents

36795

Supelco

Sethoxydim

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H29NO3S
CAS Number:
Molecular Weight:
327.48
Beilstein:
8222943
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

−20°C

SMILES string

CCC\C(=N/OCC)C1=C(O)CC(CC(C)SCC)CC1=O

InChI

1S/C17H29NO3S/c1-5-8-14(18-21-6-2)17-15(19)10-13(11-16(17)20)9-12(4)22-7-3/h12-13,19H,5-11H2,1-4H3/b18-14+

InChI key

CSPPKDPQLUUTND-NBVRZTHBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sethoxydim is a grass-active herbicide that is very effective for postemergence control of annual and perennial grasses. Sethoxydim belongs to the cyclohexanediones family and is mainly applied to broad-leaved crops, including cotton, oilseed rape, soybeans, sugar beets, fodder beets, sunflowers, spinach, potatoes, tobacco, peanuts, strawberries, alfalfa, flax, and vegetables. The herbicide act by inhibiting plastid acetyl-CoA carboxylase (ACCase) of monocotyledonous species, causing chloroplast bleaching, tissue necrosis, growth reduction, and ultimately death of a plant.

Sethoxydim is not approved for its use in the European Union. Maximum residue levels (MRLs) have been set according to Reg (EC) No 839/2008 for sethoxydim (to be calculated as the sum of sethoxydim and clethodim including degradation products) for various products of plant and animal origin from 0.05 to 1 mg/kg.

Application

The analytical standard can be used to:

  • Study the inhibition effect of acetyl-CoA carboxylase in barley chloroplasts using cycloxydim, sethoxydim, and clethodim herbicides
  • Study the photochemical changes and oxidative damage in four foxtail millet varieties after exposure to sethoxydim
  • Study the resistance mechanism of chloroplastic acetyl-CoA carboxylase from green foxtail (Setaria viridis L. Beauv.) towards cyclohexanedione herbicide sethoxydim
  • Study the inhibition mechanism of acetyl-CoA carboxylase by sethoxydim and haloxyfop using a semipurified enzyme preparation extracted from black Mexican sweet maize
  • Evaluate the tolerance of 11 grass species used in minimal-to-no-mow turf areas to three postemergence herbicides applied at one- and twofold recommended label rates
  • Generate sethoxydim-resistant mutants of Micractinium sp. by UV-C mutagenesis for increased cellular triacylglycerol (TAG) without compromising on growth rate

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Notes: Inhibition of the Acetyl-CoA Carboxylase of Barley Chloroplasts by Cycloxydim and Sethoxydim
Focke M and Lichtenthaler HK
Zeitschrift fur Naturforschung C, 11-12 null
Jinjin Diao et al.
Metabolic engineering, 51, 88-98 (2018-11-06)
Dietary omega-3 long-chain polyunsaturated fatty acids docosahexaenoic acid (DHA, C22:6) can be synthesized in microalgae Crypthecodinium cohnii; however, its productivity is still low. Here, we established a new protocol termed as "chemical modulator based adaptive laboratory evolution" (CM-ALE) to enhance
S Lin et al.
Bioorganic & medicinal chemistry letters, 9(4), 551-554 (1999-03-31)
Sethoxydim, a commercially available cyclohexanedione class herbicide by targeting the enzymatic activity of acetyl-coenzyme A carboxylase, has been found to moderately inhibit the activity of 4-hydroxyphenylpyruvate dioxygenase, a key enzyme in the biosynthesis of plastoquinones and tocopherols in plants.
B S Pakdaman et al.
Pakistan journal of biological sciences : PJBS, 10(15), 2480-2484 (2007-08-01)
The fungicidal effect of sethoxydim on the canola (Brassica napus var. Olifera) white stem rot pathogen (Sclerotinia sclerotiorum) encouraged us to conduct a series of studies on the mechanism of the antifungal activity of this herbicide commonly applied in Iranian
B J Incledon et al.
Journal of agricultural and food chemistry, 47(1), 299-304 (1999-11-24)
Acetyl-coenzyme A carboxylase (ACCase) isozymes were separated from cyclohexanedione-resistant and -susceptible maize. ACCase240 from resistant maize was 3.7-, >77-, and 12.8-fold more resistant to inhibition by clethodim, sethoxydim, and tralkoxydim, respectively, than ACCase240 from susceptible maize. The resistant ACCase240 preparation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service