Skip to Content
Merck
All Photos(2)

Key Documents

35352

Supelco

Dioxabenzofos

PESTANAL®, analytical standard

Synonym(s):

(RS)-2-Methoxy-4H-1,3,2δ5-benzodioxaphosphinine 2-sulfide, (RS)-2-Methoxy-4H-1,3,2δ5-benzodioxaphosphorine 2-sulfide, 2-Methoxy-4H-1,3,2-benzodioxaphosphorin 2-sulfide, Dioxabenzophos, Salithion

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9O3PS
CAS Number:
Molecular Weight:
216.19
Beilstein:
1875270
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

COP1(=S)OCc2ccccc2O1

InChI

1S/C8H9O3PS/c1-9-12(13)10-6-7-4-2-3-5-8(7)11-12/h2-5H,6H2,1H3

InChI key

OUNSASXJZHBGAI-UHFFFAOYSA-N

Application

Dioxabenzofos may be used as an analytical reference standard for the determination of the analyte in food, animal tissues, Oolong tea and vegetables/fruits by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiresidue analysis of pesticides in vegetables and fruits using a high capacity absorbent polymer for water.
Obana H, et al.
Analyst, 124(8), 1159-1165 (1999)
Multiresidue method for the determination of pesticides in Oolong tea using QuEChERS by gas chromatography-triple quadrupole tandem mass spectrometry.
Wu CC.
Food Chemistry, 229(1), 580-587 (2017)
Shanshan Zhou et al.
Chirality, 21(10), 922-928 (2009-01-24)
Enantioseletive toxicities of chiral pesticides have become an environmental concern recently. In this study, we evaluated the enantiomeric separation of salithion on a suite of commercial chiral columns and assessed the toxicity of enantiomers toward butyrylcholinesterase and Daphnia magna. Satisfactory
M Eto
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 18(1), 119-145 (1983-01-01)
Chemical and biochemical studies on three types of cyclic phosphoryl compounds are discussed in connection with the development of insecticides. They are five-membered cyclic phosphoramidates (I), six-membered cyclic phosphates (II) and bridged bicyclic phosphates (III). Development of I has started
Rapid method for the determination of 180 pesticide residues in foods by gas chromatography/mass spectrometry and flame photometric detection.
Okihashi M, et al.
Journal of Pesticide Science, 30(4), 368-377 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service