Skip to Content
Merck
All Photos(1)

Key Documents

34750

Sigma-Aldrich

2,6-Di-tert-butyl-4-methylphenol

purum, ≥99.0% (GC)

Synonym(s):

2,6-Di-tert-butyl-p-cresol, BHT, Butylated hydroxytoluene, Butylhydroxytoluene, DBPC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]2C6H2(CH3)OH
CAS Number:
Molecular Weight:
220.35
Beilstein:
1911640
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

7.6 (vs air)

Quality Level

vapor pressure

<0.01 mmHg ( 20 °C)

grade

purum

Assay

≥99.0% (GC)

autoignition temp.

878 °F

bp

265 °C (lit.)

mp

68-72 °C
69-73 °C (lit.)

solubility

methanol: 0.1 g/mL, clear, colorless
methanol: soluble 100 mg/mL, clear, colorless

SMILES string

Cc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C15H24O/c1-10-8-11(14(2,3)4)13(16)12(9-10)15(5,6)7/h8-9,16H,1-7H3

InChI key

NLZUEZXRPGMBCV-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

2,6-Di-tert-butyl-4-methylphenol is an 4-alkylphenol. It is an antioxidant and exhibit toxicities mediated by oxidative metabolism to electrophilic quinone methides. It acts as Michael acceptor and its reaction with simple nucleophiles and proteins have been reported. Reaction of 2,6-di-tert-butyl-4-methylphenol with ytterbium(II)-benzophenone dianion complex has been reported. 2,6-Di-tert-butyl-4-methylphenol, a food additive, promotes acute pulmonary toxicity and tumor growth in mice.

Application

2,6-Di-tert-butyl-4-methylphenol may be used in the preparation of an organoaluminum compound, methylaluminum bis(2,6-di-tert-butyl-4-alkylphenoxide).

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

260.6 °F - open cup

Flash Point(C)

127 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Lewis et al.
Chemical research in toxicology, 9(8), 1368-1374 (1996-12-01)
4-Alkylphenols, such as the antioxidant 2, 6-di-tert-butyl-4-methylphenol (BHT), exhibit toxicities that appear to be mediated by their oxidative metabolism to electrophilic quinone methides. Reactions of these Michael acceptors with simple nucleophiles and proteins have been reported, but little information is
Methylaluminum bis (2, 6-di-tert-butyl-4-alkylphenoxide). A new reagent for obtaining unusual equatorial and anti-Cram selectivity in carbonyl alkylation.
Maruoka K, et al.
Journal of the American Chemical Society, 107(15), 4573-4576 (1985)
Novel crystal structure of ytterbium (II)-benzophenone dianion complex and its reaction with 2, 6-di-tert-butyl-4-methylphenol.
Hou Z, et al.
Journal of the Chemical Society. Chemical Communications, 9, 722-724 (1999)
Rene Kupfer et al.
Chemical research in toxicology, 15(8), 1106-1112 (2002-08-20)
Acute pulmonary toxicity and tumor promotion by the food additive 2,6-di-tert-butyl-4-methylphenol (BHT) in mice are well documented. These effects have been attributed to either of two quinone methides, 2,6-di-tert-butyl-4-methylenecyclohexa-2,5-dienone (BHT-QM) formed through direct oxidation of BHT by pulmonary cytochrome P450
A M Vijesh et al.
European journal of medicinal chemistry, 46(11), 5591-5597 (2011-10-05)
In the present study two new series of Hantzsch 1,4-dihydropyridine derivatives (1,4-DHPs) containing substituted pyrazole moiety (4a-f and 5a-f) were synthesized by the reaction of 3-aryl-1H-pyrazole-4-carbaldehydes with 1,3-dicarbonylcompounds (ethylacetoacetate and methylacetoacetate) and ammonium acetate. The newly synthesized compounds were characterized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service