Skip to Content
Merck
All Photos(1)

Documents

34134

Supelco

B-Trichothecene mix, (DON, NIV, 3-AcDON, 15-AcDON)

~100 μg/mL in acetonitrile (each of DON, NIV, 3-AcDON and 15-AcDON), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

~100 μg/mL in acetonitrile (each of DON, NIV, 3-AcDON and 15-AcDON)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

multi-component solution

storage temp.

−20°C

General description

Trichothecene mycotoxins are the largest group of toxins produced by moulds of genus Fusarium. Trichothecenes are divided into four types (A-D) according to their characteristic functional groups. The epidemiological surveys have revealed that A and B of trichothecenes are widely distributed in cereals and feed as natural pollutants. The most predominant type-B Trichothecenes are Deoxynivalenol (DON), nivalenol (NIV), 3-acetyldeoxynivalenol (3-AcDON) and 15- acetyldeoxynivalenol (15-AcDON).

Application

B-trichothecene mix may be used as reference standard in quantitative determination of type B trichothecenes using coupling liquid chromatography with tandem mass spectrometry (LC/MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

purity : ≥97.0% (HPLC) for all components

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tomoyuki Kadota et al.
Rapid communications in mass spectrometry : RCM, 25(23), 3481-3490 (2011-11-19)
A method coupling liquid chromatography with tandem mass spectrometry (LC/MS/MS) was developed for the simultaneous quantitative determination of trichothecenes, nivalenol, deoxynivalenol, deoxynivalenol-3-glucoside, fusarenon-X, 3-acetyldeoxynivalenol, 15-acetyldeoxynivalenol, isotrichodermin, calonectrin, 3-deacetylcalonectrin, 15-deacetylcalonectrin, 3,15-diacetylnivalenol, 4,15-diacetylnivalenol, 3,15-diacetyldeoxynivalenol, and 3,4,15-triacetylnivalenol. The analytical parameters of trichothecenes and
R Krska et al.
Fresenius' journal of analytical chemistry, 371(3), 285-299 (2001-11-02)
The aim of this review is to describe the state-of-the-art in the analysis of A- and B-trichothecene mycotoxins in cereals and to support knowledge and experience exchange between laboratories in the field of Fusarium mycotoxin analysis. Current screening tests and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service