Skip to Content
Merck
All Photos(2)

Key Documents

32552

Supelco

Decoquinate-d5

VETRANAL®, analytical standard

Synonym(s):

Ethyl 6-decyloxy-7-ethoxy-d5-4-hydroxy-3-quinolinecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24D5H30NO5
CAS Number:
Molecular Weight:
422.57
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

mass shift

M+5

storage temp.

2-8°C

SMILES string

[2H]C([2H])([2H])C([2H])([2H])Oc1cc2ncc(C(=O)OCC)c(O)c2cc1OCCCCCCCCCC

Application

Decoquinate-d5 may be used as an analytical standard for the determination of decoquinate in chicken liver samples, bovine milk samples, animal feed samples and egg samples by liquid chromatography-tandem mass spectrometry (LC-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Multi-residue confirmatory method for the determination of twelve coccidiostats in chicken liver using liquid chromatography tandem mass spectrometry.
Olejnik M, et al.
Journal of Chromatography A, 1216(46), 8141-8148 (2009)
Multiresidue method for the simultaneous determination of veterinary medicinal products, feed additives and illegal dyes in eggs using liquid chromatography-tandem mass spectrometry.
Piatkowska M, et al.
Food Chemistry, 197(1-2), 571-580 (2016)
Determination of eleven coccidiostats in animal feed by liquid chromatography-tandem mass spectrometry at cross contamination levels.
Cronly M, et al.
Analytica Chimica Acta, 700(1-2), 26-33 (2011)
Development and validation of a liquid chromatographic-tandem mass spectrometric method for determination of eleven coccidiostats in milk.
Nasz S, et al.
Food Chemistry, 133(2), 536-543 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service