Skip to Content
Merck
All Photos(1)

Key Documents

31493

Sigma-Aldrich

Sodium salicylate

puriss. p.a., reag. Ph. Eur., 99.5-101.0% (calc. to the dried substance)

Synonym(s):

2-Hydroxybenzoic acid sodium salt, Salicylic acid sodium salt, Sodium 2-hydroxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4COONa
CAS Number:
Molecular Weight:
160.10
Beilstein:
3732792
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
reag. Ph. Eur.

Quality Level

grade

puriss. p.a.

Assay

99.5-101.0% (calc. to the dried substance)

impurities

acidic reac. impurities, complies
sulfite or thiosulfate, complies
≤0.001% heavy metals (as Pb)

loss

≤0.3% loss on drying, 105 °C

mp

>300 °C (lit.)

anion traces

chloride (Cl-): ≤20 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

Fe: ≤10 mg/kg

suitability

in accordance for appearance of solution

SMILES string

[Na+].Oc1ccccc1C([O-])=O

InChI

1S/C7H6O3.Na/c8-6-4-2-1-3-5(6)7(9)10;/h1-4,8H,(H,9,10);/q;+1/p-1

InChI key

ABBQHOQBGMUPJH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium salicylate can be prepared from the reaction of salicylic acid and ammonia. It exhibits fluorescence at higher concentrations and effectively absorbs β-particles. Due to these properties, it has been employed as fluor for the radioactivity detection in various gels.

Application

Sodium salicylate may be used as phenolic reagent in salicylate method for the determination of ammonia. It may be employed as a hydrotropic agent to study the release of anticancer drug camptothecin (CPT) from the mixed micelles [composed of Pluronic P105 (P105) and D-α-tocopheryl polyethylene glycol 1000 succinate (TPGS)].

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

208.9 °F - Pensky-Martens closed cup

Flash Point(C)

98.3 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yan Gao et al.
Colloids and surfaces. B, Biointerfaces, 64(2), 194-199 (2008-03-08)
To increase the solubility and cytotoxicity of poorly soluble anticancer drug camptothecin (CPT), mixed micelles made of Pluronic P105 (P105) and d-alpha-tocopheryl polyethylene glycol 1,000 succinate (TPGS) were prepared. The interaction of Pluronic and TPGS was studied and critical micelle
Ammonia determination based on indophenol formation with sodium salicylate.
Verdouw H, et al.
Water Research, 12(6), 399-402 (1978)
Fluorographic detection of radioactivity in polyacrylamide gels with the water-soluble fluor, sodium salicylate.
J P Chamberlain
Analytical biochemistry, 98(1), 132-135 (1979-09-15)
Kui Wang et al.
Journal of medicinal chemistry, 52(20), 6402-6412 (2009-10-16)
Viologens are showing an increasing number of scientific and technical applications in addition to their use as herbicides. However, their high toxicity poses considerable risks to human health, society, and the environment. In this context, we propose a new therapeutic
Yan-Yan Su et al.
PloS one, 7(10), e46969-e46969 (2012-10-17)
Sodium salicylate (NaSal), an aspirin metabolite, can cause tinnitus in animals and human subjects. To explore neural mechanisms underlying salicylate-induced tinnitus, we examined effects of NaSal on neural activities of the medial geniculate body (MGB), an auditory thalamic nucleus that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service