Skip to Content
Merck
All Photos(1)

Key Documents

26454

Sigma-Aldrich

Magnesium stearate

puriss., meets analytical specification of Ph. Eur., BP, ≥90% stearic and palmitic acid basis, ≥40% stearic acid basis (GC), 4.0-5.0% Mg basis (calc on dry sub.)

Synonym(s):

Stearic acid magnesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)16CO2]2Mg
CAS Number:
Molecular Weight:
591.24
Beilstein:
3919702
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

grade

puriss.

Assay

≥40% stearic acid basis (GC)
≥90% stearic and palmitic acid basis
4.0-5.0% Mg basis (calc on dry sub.)

quality

meets analytical specification of Ph. Eur., BP

impurities

acidity or alkalinity, complies
microbiological impurity, in accordance
residual solvents, complies

loss

≤6.0% loss on drying, 105 °C

mp

200 °C (lit.)

acid value

195‑210(fatty acid)

solubility

alcohol: insoluble
diethyl ether: insoluble
water: insoluble

anion traces

chloride (Cl-): ≤1000 mg/kg
sulfate (SO42-): ≤5000 mg/kg

cation traces

Cd: ≤3 mg/kg
Ni: ≤5 mg/kg
Pb: ≤10 mg/kg

suitability

in accordance for fatty acid composition
passes test for identity

application(s)

pharmaceutical (small molecule)

SMILES string

CCCCCCCCCCCCCCCCCC(=O)O[Mg]OC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/2C18H36O2.Mg/c2*1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h2*2-17H2,1H3,(H,19,20);/q;;+2/p-2

InChI key

HQKMJHAJHXVSDF-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Magnesium stearate has been used as a tablet and capsule lubricant. It has also been used for preparing microcapsules.

Biochem/physiol Actions

Magnesium stearate is the magnesium salt of stearic acid that possess lubricating properties and hence prevents ingredients from sticking to manufacturing equipment during the compression of chemical powders into solid tablets. Dry coating of drugs with magnesium stearate leads to flow improvement, flow-aid and lubrication effects, tabletability as well as non-inhibited dissolution rate.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M J Amorim et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 52(1), 39-44 (2001-07-05)
Several different peptides and proteins, such as the pancreatic trypsin inhibitor, growth factors and trefoil peptides, are known to play important roles in maintaining the structure and function of the gastrointestinal wall. With the advent of recombinant biotechnology, it has
<BIG>Marleen de Veij</BIG>
Journal of Raman Spectroscopy, 40 (2008)
Ziyaur Rahman et al.
International journal of pharmaceutics, 475(1-2), 462-470 (2014-09-01)
Clinical performance of an amorphous solid dispersion (ASD) drug product is related to the amorphous drug content because of the greater bioavailability of this form of the drug than its crystalline form. Therefore, it is paramount to monitor the amorphous
A Kluk et al.
International journal of pharmaceutics, 485(1-2), 1-6 (2015-03-05)
Minitablets are a novel, multi-compartment solid drug formulation, particularly intended for children between 1 and 6 years of age. Available literature shows that even infants are capable of swallowing a single minitablet. In this study, we have explored the level
Julia Grund et al.
International journal of pharmaceutics, 469(1), 94-101 (2014-04-22)
The objective of this study was to identify and evaluate key polymer properties affecting direct compression and drug release from water-insoluble matrices. Commonly used polymers, such as Kollidon(®) SR, Eudragit(®) RS and ethyl cellulose, were characterized, formulated into tablets and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service