Skip to Content
Merck
All Photos(1)

Key Documents

25666

Supelco

Chloroform – isoamyl alcohol mixture

24:1, ≥99.5%, for molecular biology, BioUltra

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
ND.02

product name

Chloroform – isoamyl alcohol mixture, BioUltra, for molecular biology, 24:1, ≥99.5% (chloroform + isoamyl alcohol, GC)

grade

for molecular biology

Quality Level

product line

BioUltra

Assay

≥99.5% (chloroform + isoamyl alcohol, GC)

form

liquid

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected
≤0.001% free alkali (as NH3)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.15
λ: 280 nm Amax: 0.1

SMILES string

ClC(Cl)Cl.CC(C)CCO

InChI

1S/C5H12O.CHCl3/c1-5(2)3-4-6;2-1(3)4/h5-6H,3-4H2,1-2H3;1H

InChI key

BKHZIBWEHPHYAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Components

mixture of chloroform and isoamyl alcohol in the ratio of 24:1 (v/v)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justine Cheval et al.
mSphere, 4(3) (2019-06-07)
The use of high-throughput sequencing (HTS) to identify viruses in biologicals differs from current molecular approaches, since its use enables an unbiased approach to detection without the need to design specific primers to preamplify target sequences. Its broad range of
Iris E M de Jong et al.
Hepatology (Baltimore, Md.), 69(4), 1719-1734 (2018-12-07)
Peribiliary glands (PBG) are a source of stem/progenitor cells organized in a cellular network encircling large bile ducts. Severe cholangiopathy with loss of luminal biliary epithelium has been proposed to activate PBG, resulting in cell proliferation and differentiation to restore
Montserrat Saladié et al.
Frontiers in microbiology, 11, 572504-572504 (2020-10-31)
In recent years the study of the commensal microbiota is driving a remarkable paradigm shift in our understanding of human physiology. However, intrinsic technical difficulties associated with investigating the Microbiomics of some body niches are hampering the development of new
Simon C Johnson et al.
Kidney international, 95(2), 455-466 (2018-11-26)
Mitochondrial diseases represent a significant clinical challenge. Substantial efforts have been devoted to identifying therapeutic strategies for mitochondrial disorders, but effective interventions have remained elusive. Recently, we reported attenuation of disease in a mouse model of the human mitochondrial disease

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service