Key Documents
24531
Hamilton® syringe, luer tip
702LT, volume 25 μL, needle size (not included)
About This Item
UNSPSC Code:
41122001
material
glass body
needle size
(not included)
feature
without needle Luer tip
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 80401
needle L
, n/a
volume
25 μL
compatibility
for use with Hamilton 702LT (Luer tip)
General description
Hamilton syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed. The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques null
Pharmacology for the Prehospital Professional null
Guy, Jeffrey S.
Pharmacology for the Prehospital Professional, 54-54 (2009)
Wolf, Don P.
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 192-192 (2012)