Skip to Content
Merck
All Photos(3)

Key Documents

240664

Sigma-Aldrich

1,2-Dichlorobenzene

anhydrous, 99%

Synonym(s):

o-DCB, o-dichlorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4Cl2
CAS Number:
Molecular Weight:
147.00
Beilstein:
606078
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous

Quality Level

vapor density

5.1 (vs air)

vapor pressure

1.2 mmHg ( 20 °C)
1.6 mmHg ( 35 °C)

Assay

99%

form

liquid

autoignition temp.

1198 °F

expl. lim.

9.2 %

impurities

<0.003% water
<0.005% water (100 mL pkg)

evapn. residue

<0.0003%

refractive index

n20/D 1.551 (lit.)

bp

178-180 °C (lit.)

mp

−18-−17 °C (lit.)

density

1.306 g/mL at 25 °C (lit.)

SMILES string

Clc1ccccc1Cl

InChI

1S/C6H4Cl2/c7-5-3-1-2-4-6(5)8/h1-4H

InChI key

RFFLAFLAYFXFSW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,2-Dichlorobenzene may be used as a starting material in the synthesis of (phenyl)[2-(trimethylsilyl)phenyl]iodonium triflates and 1,2-dibutylbenzene.

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Da Young Hwang et al.
Nanotechnology, 28(2), 025603-025603 (2016-12-08)
We report that a high local strain was obtained for multilayer MoS2 nanoscrolls decorated with noble nanoparticles (Ag and Au NPs) using a rolling process beyond breaking or slipping of MoS2. The local strain was estimated through the bending strain in the nanoscrolls and
Critical light instability in CB/DIO processed PBDTTT-EFT: PC 71 BM organic photovoltaic devices.
Pearson AJ, et al.
Organic Electronics, 30, 225-236 (2016)
(Phenyl)[2?(Trimethylsilyl) Phenyl] Iodonium Triflate. An Efficient And Mild Benzyne Precursor
Kitamura T, et al.
Organic Syntheses, 78, 104-104 (2002)
Oxygen reduction at a water-1, 2-dichlorobenzene interface catalyzed by cobalt tetraphenyl porphyrine-A fuel cell approach.
Peljo P, et al.
International Journal of Hydrogen Energy, 36(16), 10033-10043 (2011)
Phosphine?Nickel Complex Catalyzed Cross?Coupling of Grignard Reagents with Aryl and Alkenyl Halides: 1, 2?Dibutylbenzene.
Kumada, M, et al.
Organic Syntheses, 58, 127-127 (1978)

Articles

Single molecule electronics is the endeavour of constructing electronic circuitry with single molecules as the fundamental building block.

Single molecule electronics is the endeavour of constructing electronic circuitry with single molecules as the fundamental building block.

Single molecule electronics is the endeavour of constructing electronic circuitry with single molecules as the fundamental building block.

Single molecule electronics is the endeavour of constructing electronic circuitry with single molecules as the fundamental building block.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service