Skip to Content
Merck
All Photos(1)

Key Documents

22060

Supelco

(−)-Carvone

analytical standard

Synonym(s):

(−)-p-Mentha 6,8-diene 2-one, (R)-5-Isopropenyl-2-methyl-2-cyclohexenone, Carvol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
Beilstein:
2206714
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (sum of enantiomers, GC)

optical activity

[α]20/D −61.5±2°, neat

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.498

bp

228-230 °C (lit.)

density

0.960 g/mL at 20 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(=C)[C@@H]1CC=C(C)C(=O)C1

InChI

1S/C10H14O/c1-7(2)9-5-4-8(3)10(11)6-9/h4,9H,1,5-6H2,2-3H3/t9-/m1/s1

InChI key

ULDHMXUKGWMISQ-SECBINFHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(-)-Carvone, a monoterpene ketone which is the main active component of mentha plant species like Mentha spicata. It has antinociceptive activity which is found to be associated with decreased peripheral nerve excitability.

Application

It has been used as reference standard in TLC analysis to detect the presence of ketonic monoterpene in Aloysia species.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: mentha zingiber

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antispasmodic effects of Aloysia polystachya and A. gratissima tinctures and extracts are due to non-competitive inhibition of intestinal contractility induced by acethylcholine and calcium.
Consolini, Alicia E., et al.
Revista Brasileira de Farmacognosia, 21.5, 889-900 (2011)
Antinociceptive activity of (-)-carvone: evidence of association with decreased peripheral nerve excitability.
Goncalves, Juan Carlos Ramos, et al.
Biological & Pharmaceutical Bulletin, 31.5, 1017-1020 (2008)
Burak Tepe et al.
Cell reports, 25(10), 2689-2703 (2018-12-06)
Cellular heterogeneity within the mammalian brain poses a challenge toward understanding its complex functions. Within the olfactory bulb, odor information is processed by subtypes of inhibitory interneurons whose heterogeneity and functionality are influenced by ongoing adult neurogenesis. To investigate this
Delphine Hardy et al.
Scientific reports, 8(1), 9385-9385 (2018-06-22)
The adult mouse olfactory bulb is continuously supplied with new neurons that mostly differentiate into granule cells (GCs). Different subtypes of adult-born GCs have been identified, but their maturational profiles and their roles in bulbar network functioning and odor behavior
Mengyang Xuan et al.
Organic letters, 14(21), 5492-5495 (2012-10-26)
The total synthesis of the cAMP signaling pathway activator (-)-alotaketal A is reported. A convergent approach to the unusual alotane sesterterpenoid skeleton was employed, exploiting a remarkable LiDBB-mediated coupling of an (R)-carvone-derived δ-lactone with an allyl bromide side chain, followed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service