Skip to Content
Merck
All Photos(1)

Key Documents

06105

Millipore

Bile esculin azide agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Enterococcus Selective Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15.0 g/L
beef extract, 5.0 g/L
casein enzymic hydrolysate, 17.0 g/L
esculin, 1.0 g/L
ferric ammonium citrate, 0.5 g/L
oxgall, 10.0 g/L
proteose peptone, 3.0 g/L
sodium azide, 0.15 g/L
sodium chloride, 5.0 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

clinical testing
environmental
food and beverages
water monitoring

microbiology

suitability

selective and differential for Enterococcus spp.
selective and differential for Streptococcus spp.

General description

Bile esculin agar (BEA) is a differential and selective medium and is mainly used to discern group D Streptococci and Enterococci based on the organism′s potential to hydrolyze esculin. BEA comprises of oxygall (bile salts, first selective ingredients) and azide (second selective ingredients), former inhibits the growth of gram positive organisms and latter inhibits the growth of gram negative organisms. BEA also comprises of nutrients ferric citrate and esculin, where esculin is a differential ingredient and is also a fluorescent compound, upon hydrolyzation the fluorescence is lost. BEA is used for bile esculin test which is based on the hydrolysis of esculin into esculetin (6, 7-dihydroxy-coumarin) and glucose by a micro-organism that produce an enzyme esculinase.

Application

Bile Esculin Azide Agar is a selective medium used for isolation and presumptive identification of faecal Streptococci.

Preparation Note

Suspend 56.65g in 1 l distilled water. Heat to boiling to dissolve the medium completely. Sterilize by autoclaving at 121°C for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of bile-esculin agar for rapid differentiation of Enterobacteriaceae.
Lindell, SHIRLEY S., and P. A. T. R. I. C. I. A. Quinn.
Journal of Clinical Microbiology, 1(5) , 440-443 (1975)

Articles

Streptococci- Overview of Detection, Identification, Differentiation and Cultivation Techniques

Streptococci- Overview of Detection, Identification, Differentiation and Cultivation Techniques

Streptococci- Overview of Detection, Identification, Differentiation and Cultivation Techniques

Streptococci- Overview of Detection, Identification, Differentiation and Cultivation Techniques

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service