Skip to Content
Merck
All Photos(1)

Key Documents

05566

Supelco

D-(+)-Glucuronic acid γ-lactone

analytical standard

Synonym(s):

D-(+)-Glucurono-6,3-lactone, D-Glucurone, D-Glucurono-6,3-lactone, Glucuronolactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
Beilstein:
83595
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (sum of anomers, GC)

optical activity

[α]/D +18.8±2°, c = 8% in H2O (after 1 hr)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

mp

172-175 °C (lit.)

application(s)

clinical testing

format

neat

SMILES string

O=C([C@@H]([C@@H](O1)[C@H](O)[C@H](O)C1=O)O)[H]

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h1-5,8-10H/t2-,3+,4-,5+/m0/s1

InChI key

UYUXSRADSPPKRZ-SKNVOMKLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

D-Glucuronic acid γ-lactone is a normal human metabolite produced from glucose as starting material. It serves as a precursor for the synthesis of a number of deoxy sugars.

Application

D-Glucuronic acid γ-lactone has been used as an analytical standard for the analysis of D-glucuronic acid γ-lactone as a component of microbial extracellular polysaccharides in biofilms by high performance liquid chromatography (HPLC) with pulsed amperometric detection (PAD) as well as diode array detection (DAD). D-(+)-Glucuronic acid γ-lactone has been used as a reference standard for the determination of D-(+)-glucuronic acid γ-lactone in energy drinks by ultra-high performance liquid chromatography-triple quadrupole
tandem mass spectrometry (UHPLC-MS/MS).

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of taurine, glucuronolactone and glucuronic acid in energy drinks by ultra high performance liquid chromatography-tandem mass spectrometry (triple quadrupole).
Ricciutelli M, et al.
Journal of Chromatography A, 1364, 303-307 (2014)
Convenient syntheses of deoxypyranose sugars from glucuronolactone.
Stanford D and Stachulski, AV
Tetrahedron Letters, 48(13), 2361-2364 (2007)
Analysis of microbial extracellular polysaccharides in biofilms by HPLC. Part I: development of the analytical method using two complementary stationary phases.
Meisen S, et al.
Analytical and Bioanalytical Chemistry, 391(3), 993-993 (2008)
Clotilde d'Errico et al.
Biotechnology and bioengineering, 112(5), 914-922 (2014-11-27)
Lignin-carbohydrate complexes (LCCs) are believed to influence the recalcitrance of lignocellulosic plant material preventing optimal utilization of biomass in e.g. forestry, feed and biofuel applications. The recently emerged carbohydrate esterase (CE) 15 family of glucuronoyl esterases (GEs) has been proposed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service