Skip to Content
Merck
All Photos(2)

Key Documents

05238

Supelco

Erythromycin A enol ether

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H65NO12
CAS Number:
Molecular Weight:
715.91
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H](C3O)N(C)C)[C@@]4(C)CC(C)=C(O4)[C@H](C)[C@@H](O)[C@]1(C)O

InChI

1S/C37H65NO12/c1-14-25-37(10,43)30(40)20(4)28-18(2)16-36(9,50-28)32(49-34-27(39)24(38(11)12)15-19(3)45-34)21(5)29(22(6)33(42)47-25)48-26-17-35(8,44-13)31(41)23(7)46-26/h19-27,29-32,34,39-41,43H,14-17H2,1-13H3/t19-,20+,21+,22-,23+,24+,25-,26+,27-,29+,30-,31+,32-,34+,35-,36-,37-/m1/s1

InChI key

JFVYXJKGJMUGRG-KJPZRSJGSA-N

General description

Erythromycin A enol ether (EAEN) is an acidic degradation product of erythromycin A (EA) which is grouped under the macrolide antibiotic class of compounds.

Application

EAEN has been used for the analysis of EAEN as degradation product of EA in honey by ultra-performance liquid chromatography equipped with tandem quadrupole mass spectrometer (UPLC-MS/MS) with positive electrospray ionisation (ESI) operating on multiple reaction monitoring (MRM) mode and in rat′s blood plasma, serum and human urine by high performance liquid chromatography-coulometric assay. It may be used as a reference standard for the detection of EAEN in erythromycin fractions by isocratic LC with evaporative light scattering detection (ELSD) and ion trap MS and in commercial erythromycin samples by LC-MS/MS.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Yamashita et al.
Journal of chromatography, 616(1), 144-150 (1993-06-23)
A sensitive high-performance liquid chromatographic (HPLC) method using column switching is described for the determination of EM523 (I), a new erythromycin derivative, in human plasma and urine. The analyte was extracted from alkalinized plasma or urine with a mixture of
Determination of erythromycin and related substances in commercial samples using liquid chromatography/ion trap mass spectrometry.
Deubel A, et al.
Journal of Chromatography A, 1136(1), 39-47 (2006)
Y Shiba et al.
Gastroenterology, 110(1), 241-250 (1996-01-01)
EM523, a nonpeptide motilin agonist, is being developed for clinical use to improve delayed gastric emptying. The aim of this study was to examine the effect of EM523 on endogenous release of gut and pancreatic hormones in conscious dogs. Motility
Identification of impurities in erythromycin by liquid chromatography-mass spectrometric detection.
Chitneni SK, et al.
Journal of Chromatography A, 1056(1-2), 111-120 (2004)
T L Peeters et al.
Biochemical and biophysical research communications, 198(2), 411-416 (1994-01-28)
Studies on the physiological role of motilin, and more recently, on the relationship between motilin and erythromycin A, have been hampered by the lack of antagonists. We now have discovered such a compound. ANQ-11125 displaces motilin bound to an homogenate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service