Skip to Content
Merck
All Photos(1)

Key Documents

04365

Sigma-Aldrich

1-Ethyl-3-methylimidazolium tetrafluoroborate

≥97.0% (HPLC/T)

Synonym(s):

EMIMBF4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11BF4N2
CAS Number:
Molecular Weight:
197.97
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (HPLC/T)

form

liquid

refractive index

n20/D 1.41
n20/D 1.413 (lit.)

bp

>350 °C (lit.)

mp

15 °C (lit.)

density

1.294 g/mL at 25 °C (lit.)

SMILES string

F[B-](F)(F)F.CCn1cc[n+](C)c1

InChI

1S/C6H11N2.BF4/c1-3-8-5-4-7(2)6-8;2-1(3,4)5/h4-6H,3H2,1-2H3;/q+1;-1

InChI key

CUNYTRQQXKCRTJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Ethyl-3-methylimidazolium tetrafluoroborate is a room temperature ionic liquid (RTIL).

Application

The enzymatic resolution of homophenylalanine ester to form enantiomeric homophenylalanine can be performed using 1-ethyl-3-methylimidazolium tetrafluoroborate as a reaction medium.

Other Notes

Air- and water-stable liquid electrolyte (m.p. 15°C)

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solubility of carbon dioxide in 1-ethyl-3-methylimidazolium tetrafluoroborate
Soriano, Allan N., Bonifacio T. Doma Jr, and Meng-Hui Li
Journal of Chemical and Engineering Data, 53.11, 2550-2555 (2008)
Air and water stable 1-ethyl-3-methylimidazolium based ionic liquids.
J.S. Wilkes et al.
Journal of the Chemical Society. Chemical Communications, 13, 965-965 (1992)
Efficient, halide free synthesis of new, low cost ionic liquids: 1, 3-dialkylimidazolium salts containing methyl-and ethyl-sulfate anions
Holbrey, John D., et al.
Green Chemistry, 4.5, 407-413 (2002)
Enzymatic Reactions in Ionic Liquids
Aldrich Chemfiles, 5(6) null
Crystal structures of frozen room temperature ionic liquids, 1-ethyl-3-methylimidazolium tetrafluoroborate (EMImBF 4), hexafluoroniobate (EMImNbF 6) and hexafluorotantalate (EMImTaF 6), determined by low-temperature X-ray diffraction
Matsumoto, Kazuhiko, et al.
Solid State Sciences, 8.10, 1250-1257 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service