Skip to Content
Merck
All Photos(1)

Key Documents

03202

Sigma-Aldrich

tert-Butyl ethyl ether

purum, ≥97.0% (GC)

Synonym(s):

2-Methyl-2-ethoxypropane, ETBE, Ethyltert-butyl ether, Ethyltert-butyl oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC(CH3)3
CAS Number:
Molecular Weight:
102.17
Beilstein:
1731469
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39021026
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

155 mmHg ( 25 °C)

Quality Level

grade

purum

Assay

≥97.0% (GC)

form

liquid

contains

~0.01% 2,6-Di-tert-butyl-4-methlyphenol as stabilizer

refractive index

n20/D 1.375 (lit.)
n20/D 1.376

bp

72-73 °C (lit.)

mp

−97 °C (lit.)

density

0.742 g/mL at 25 °C (lit.)

SMILES string

CCOC(C)(C)C

InChI

1S/C6H14O/c1-5-7-6(2,3)4/h5H2,1-4H3

InChI key

NUMQCACRALPSHD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

tert-Butyl ethyl ether (ETBE) is generally used as an antiknock additive in gasoline for the reduction of carbon monoxide and unburned hydrocarbons emission from automobile exhausts.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of additives on the antiknock properties and Reid vapor pressure of gasoline.
Da Silva R, et al.
Fuel: The Science and Technology of Fuel and Energy, 84(7-8), 951-959 (2005)
Experimental and modeling study of the gas-phase oxidation of methyl and ethyl tertiary butyl ethers.
Glaude PA, et al.
Combustion and Flame, 121(1-2), 345-355 (2000)
Yuzo Asano et al.
Drug and chemical toxicology, 34(3), 311-317 (2011-06-09)
Ethyl tertiary-butyl ether (ETBE) is commonly used as an oxygenated gasoline additive. In this study, the prenatal developmental toxicity of ETBE was determined in rabbits. New Zealand white rabbits were given ETBE by gavage at 100, 300, or 1,000 mg/kg/day
Zuquan Weng et al.
Industrial health, 49(3), 396-399 (2011-03-05)
To clarify the genotoxicity of ethyl tertiary butyl ether (ETBE), a gasoline additive, male and female C57BL/6 mice of Aldh2+/+ and Aldh2-/- genotypes, aged 8 wk, were exposed to 0, 500, 1,750, or 5,000 ppm ETBE for 6 h/day, 5
Ann de Peyster et al.
Toxicology letters, 190(1), 74-80 (2009-07-15)
These experiments were conducted to follow up on a report of testis seminiferous tubular degeneration in Fischer 344 rats treated with high doses of ethyl t-butyl ether (ETBE). Also, high doses of a related compound, methyl t-butyl ether (MTBE), had

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service