Skip to Content
Merck
All Photos(1)

Key Documents

00930585

Hederacoside C

primary reference standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C59H96O26
CAS Number:
Molecular Weight:
1221.38
Beilstein:
4229541
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

mp

222 °C (dec.) (lit.)

SMILES string

[H][C@]12CC=C3C4CC(C)(C)CC[C@@]4(CC[C@@]3(C)[C@]1(C)CCC5[C@](C)(CO)[C@H](CC[C@]25C)O[C@@H]6OC[C@H](O)[C@H](O)[C@H]6O[C@@H]7O[C@@H](C)[C@H](O)[C@@H](O)[C@H]7O)C(=O)O[C@@H]8O[C@H](CO[C@@H]9O[C@H](CO)[C@@H](O[C@@H]%10O[C@@H](C)[C@H](O)[C@@H](O)[C@H]%10O)[C@H](O)[C@H]9O)[C@@H](O)[C@H](O)[C@H]8O

InChI

1S/C59H96O26/c1-24-34(63)38(67)42(71)49(78-24)83-46-29(20-60)80-48(45(74)41(46)70)77-22-30-37(66)40(69)44(73)51(81-30)85-53(75)59-17-15-54(3,4)19-27(59)26-9-10-32-55(5)13-12-33(56(6,23-61)31(55)11-14-58(32,8)57(26,7)16-18-59)82-52-47(36(65)28(62)21-76-52)84-50-43(72)39(68)35(64)25(2)79-50/h9,24-25,27-52,60-74H,10-23H2,1-8H3/t24-,25-,27?,28-,29+,30+,31?,32+,33-,34-,35-,36-,37+,38+,39+,40-,41+,42+,43+,44+,45+,46+,47+,48+,49-,50-,51-,52-,55-,56-,57+,58+,59-/m0/s1

InChI key

RYHDIBJJJRNDSX-WBOYGZCJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Other Notes

This compound is commonly found in plants of the genus: hedera

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Eun-Ha Joh et al.
Phytotherapy research : PTR, 26(4), 546-551 (2011-09-20)
The stem-bark of Kalopanax pictus (KP, family Araliaceae), which contains triterpenoid saponins, has been shown to exhibit anticarcinogenic, antiinflammatory, antirheumatoid and antidiabetic activities. In a preliminary study, a KP methanol extract demonstrated acetylcholinesterase activity in vitro and memory enhancement in
S H Cho et al.
Archives of pharmacal research, 14(1), 19-24 (1991-03-01)
One new triterpenoidal saponin, saponin F(2) has been isolated from the bark of Kalopanax pictum Nakai var. typicum (Araliaceae), together with one known saponin, kizuta saponin K12 (1). On the basis of chemico-spectral evidences, the structure of 2 has been
Eun-Ha Joh et al.
Cellular immunology, 279(1), 103-108 (2012-11-03)
The stem bark of Kalopanax pictus Nakai (KP, family Araliaceae), of which main constituent is kalopanaxsaponin B, has been used for inflammation in Chinese traditional medicine. We isolated kalopanaxsaponin B from KP and investigated its anti-inflammatory effect in lipopolysaccharide (LPS)-stimulated
D H Kim et al.
Biological & pharmaceutical bulletin, 21(4), 360-365 (1998-05-20)
To investigate the relationship between the intestinal bacterial metabolism of kalopanaxsaponin B and H from Kalopanax pictus (Araliaceae), and their antidiabetic effect, kalopanaxsaponin B and H were metabolized by human intestinal microflora and the antidiabetic activity of their metabolites was
Jongwon Choi et al.
Journal of ethnopharmacology, 79(2), 199-204 (2002-01-22)
In the present study, we have attempted to elucidate the active components for rheumatoidal arthritis using chloroform (CHCl(3)), ethylacetate (EtOAc) and n-butanol (BuOH) fractions of the methanol extract (MeOH) of Kalopanax pictus. Kalopanaxsaponin-A and -I (KPS-A and -I, hederagenin monodesmoside)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service