Skip to Content
Merck
All Photos(1)

Key Documents

24-6190

Sigma-Aldrich

(±)-Propylene oxide

SAJ special grade, ≥99.0%

Synonym(s):

(±)-Methyloxirane, 1,2-Epoxypropane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O
CAS Number:
Molecular Weight:
58.08
Beilstein:
79763
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

SAJ special grade

vapor density

2 (vs air)

vapor pressure

29.43 psi ( 55 °C)
8.59 psi ( 20 °C)

Assay

≥99.0%

autoignition temp.

1378 °F

expl. lim.

37 %

availability

available only in Japan

refractive index

n20/D 1.366 (lit.)

bp

34 °C (lit.)

mp

−112 °C (lit.)

density

0.83 g/mL at 25 °C (lit.)

SMILES string

CC1CO1

InChI

1S/C3H6O/c1-3-2-4-3/h3H,2H2,1H3

InChI key

GOOHAUXETOMSMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 1 - Muta. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-36.4 °F - Equilibrium method

Flash Point(C)

-38 °C - Equilibrium method

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denis Merlet et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 209(2), 315-322 (2011-02-25)
The application of the G-SERFph pulse sequence is presented on enantiomeric mixtures dissolved in a chiral liquid crystal. It aims at editing, within one single 2D spectrum, every proton coupling which is experienced by a given proton site in the
Josep Casamada Ribot et al.
Chemical communications (Cambridge, England), 46(37), 6971-6973 (2010-08-24)
The aqueous gelation of a quaternary ammonium oligo(propylene oxide)-based ionic liquid yields ion gels with a reverse thermoresponsive behavior (i.e., mechanical moduli and viscosity increase with temperature) and enhanced ionic conductivities.
Aude Bouchet et al.
The Journal of organic chemistry, 76(10), 4178-4181 (2011-04-08)
ECD and NMR experiments show that the complexation of propylene oxide (PrO) within the cavity of an enantiopure water-soluble cryptophane 1 in NaOH solution is enantioselective and that the (R)-PrO@PP-1 diastereomer is more stable than the (S)-PrO@PP-1 diastereomer with a
Javix Thomas et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(16), 4582-4587 (2011-03-25)
Chirality recognition in the hydrogen-bonded glycidol···propylene oxide complex has been studied by using rotational spectroscopy and ab initio calculations. An extensive conformational search has been performed for this binary adduct at the MP2/6-311++G(d,p) level of theory and a total of
Sascha Brunke et al.
PLoS pathogens, 10(10), e1004478-e1004478 (2014-10-31)
Candida glabrata is one of the most common causes of candidemia, a life-threatening, systemic fungal infection, and is surpassed in frequency only by Candida albicans. Major factors contributing to the success of this opportunistic pathogen include its ability to readily

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service