Skip to Content
Merck
All Photos(1)

Key Documents

17-0090

Sigma-Aldrich

Kerosene

CP

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
15101502

grade

CP

vapor density

4.5 (vs air)

vapor pressure

0.23 mmHg ( 20 °C)

form

liquid

autoignition temp.

442 °F

expl. lim.

5 %

availability

available only in Japan

bp

190-250 °C (lit.)

density

0.8 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kent L Gee et al.
The Journal of the Acoustical Society of America, 123(6), 4082-4093 (2008-06-10)
To address the question of the role of nonlinear effects in the propagation of noise radiated by high-power jet aircraft, extensive measurements were made of the F-22A Raptor during static engine run-ups. Data were acquired at low-, intermediate-, and high-thrust
J A Namocatcat et al.
Journal of contaminant hydrology, 67(1-4), 177-194 (2003-11-11)
Evolution of trimethylbenzoic acids in the KC-135 aquifer at the former Wurtsmith Air Force Base (WAFB), Oscoda, MI was examined to determine the functionality of trimethylbenzoic acids as key metabolite signatures in the biogeochemical evolution of an aquifer contaminated with
J V Rogers et al.
Journal of applied toxicology : JAT, 21(6), 521-525 (2001-12-18)
Dermal absorption of JP-8 jet fuel can lead to skin irritation within hours after exposure. This study detected the formation of oxidative species and low-molecular-weight DNA in rat skin as potential indicators of JP-8-induced skin injury. At 0, 1, 2
Rakesh Sharma et al.
Magnetic resonance imaging, 28(7), 1030-1048 (2010-07-29)
The toxicity of jet fuels was measured using noninvasive magnetic resonance microimaging (MRM) at 900-MHz magnetic field. The hypothesis was that MRM can visualize and measure the epidermis exfoliation and hair follicle size of rat skin tissue due to toxic
Narayanasamy Kanikkannan et al.
Toxicology, 175(1-3), 35-47 (2002-06-07)
Jet A and JP-8 are the major jet fuels used in civilian and military (US Air Force) flights, respectively. JP-8+100 is a new jet fuel recently introduced by US Air Force in some of its locations. The purpose of this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service