Skip to Content
Merck
All Photos(1)

Key Documents

13-0370

Sigma-Aldrich

Heptane

SAJ first grade, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH3
CAS Number:
Molecular Weight:
100.20
Beilstein:
1730763
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

grade

SAJ first grade

vapor density

3.5 (vs air)

vapor pressure

40 mmHg ( 20 °C)
83 mmHg ( 37.7 °C)

Assay

≥98.0%

form

liquid

autoignition temp.

433 °F

expl. lim.

7 %

availability

available only in Japan

refractive index

n20/D 1.387 (lit.)

bp

98 °C (lit.)

mp

−91 °C (lit.)

density

0.684 g/mL at 25 °C (lit.)

SMILES string

CCCCCCC

InChI

1S/C7H16/c1-3-5-7-6-4-2/h3-7H2,1-2H3

InChI key

IMNFDUFMRHMDMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F

Flash Point(C)

-4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manuela Nagel et al.
Plant, cell & environment, 38(6), 1011-1022 (2014-10-21)
Globally, over 7.4 million accessions of crop seeds are stored in gene banks, and conservation of genotypic variation is pivotal for breeding. We combined genetic and biochemical approaches to obtain a broad overview of factors that influence seed storability and
Thushara Karunasekara et al.
Journal of chromatography. A, 1218(6), 809-816 (2011-01-07)
Partition coefficients for varied compounds were determined for the organic solvent-propylene carbonate biphasic partition system where the organic solvent is n-heptane, isopentyl ether or 1-octanol. These partition coefficient databases are analyzed using the solvation parameter model facilitating a quantitative comparison
Donghoon M Lee et al.
Genetics, 200(3), 863-872 (2015-05-15)
Germline cells segregate from the soma to maintain their totipotency, but the cellular mechanisms of this segregation are unclear. The Drosophila melanogaster embryo forms a posterior group of primordial germline cells (PGCs) by their division from the syncytial soma. Extended
Chris G C Jacobs et al.
Insect biochemistry and molecular biology, 60, 7-12 (2015-03-10)
Insects have been extraordinary successful in colonizing terrestrial habitats and this success is partly due to a protective cuticle that mainly contains chitin and proteins. The cuticle has been well studied in larvae and adults, but little attention has been
Bing Chen et al.
Journal of hazardous materials, 188(1-3), 369-374 (2011-02-26)
The influence of the initial fuel temperature on the burning behavior of n-heptane pool fire was experimentally studied at the State Key Laboratory of Fire Science (SKLFS) large test hall. Circular pool fires with diameters of 100mm, 141 mm, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service