Skip to Content
Merck
All Photos(1)

Key Documents

07-2080

Sigma-Aldrich

1,2-Dichloroethane

JIS special grade, ≥99.5%

Synonym(s):

Ethylene chloride, Ethylene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2Cl
CAS Number:
Molecular Weight:
98.96
Beilstein:
605264
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

grade

JIS special grade

vapor density

3.4 (20 °C, vs air)

vapor pressure

87 mmHg ( 25 °C)

Assay

≥99.5%

form

viscous liquid

autoignition temp.

775 °F

expl. lim.

15.6 %

availability

available only in Japan

refractive index

n20/D 1.444 (lit.)

bp

83 °C (lit.)

mp

−35 °C (lit.)

density

1.256 g/mL at 25 °C (lit.)

SMILES string

ClCCCl

InChI

1S/C2H4Cl2/c3-1-2-4/h1-2H2

InChI key

WSLDOOZREJYCGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

François Perreault et al.
Chemical Society reviews, 44(16), 5861-5896 (2015-03-27)
Graphene-based materials are gaining heightened attention as novel materials for environmental applications. The unique physicochemical properties of graphene, notably its exceptionally high surface area, electron mobility, thermal conductivity, and mechanical strength, can lead to novel or improved technologies to address
Rongke Gao et al.
Biosensors & bioelectronics, 72, 230-236 (2015-05-20)
We report the application of a fully automated surface-enhanced Raman scattering (SERS)-based solenoid-embedded microfluidic device to the quantitative and sensitive detection of anthrax biomarker poly-γ-D-glutamic acid (PGA) in solution. Analysis is based on the competitive reaction between PGA and PGA-conjugated
Tatiana Wolfe et al.
Nanomedicine : nanotechnology, biology, and medicine, 11(5), 1277-1283 (2015-02-06)
We report potent radiosensitization of prostate cancers in vitro and in vivo using goserelin-conjugated gold nanorods. Progressive receptor-mediated internalization of conjugated nanorods over time increases the radiation interaction cross-section of cells and contributes to the effects observed in vitro. The
Peiyu Ge et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(29), 11558-11563 (2012-06-06)
The photochemical reactivity of osmocene in a biphasic water-organic solvent system has been investigated to probe its water splitting properties. The photoreduction of aqueous protons to hydrogen under anaerobic conditions induced by osmocene dissolved in 1,2-dichloroethane and the subsequent water
Tom J Stockmann et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(47), 13206-13216 (2011-10-26)
The complexation reactions between strontium (Sr(2+)) and octyl(phenyl)-N,N-diisobutylcarbamoylmethylphosphine oxide (CMPO) were studied at the aqueous|1,2-dichloroethane (w|DCE) and aqueous|room-temperature ionic liquid (w|RTIL) microinterfaces, in order to understand its extraction in reprocessing spent nuclear fuels, remediation of environmental contamination, and potential radiological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service