Skip to Content
Merck
All Photos(1)

Key Documents

03-4450

Sigma-Aldrich

(±)-1,3-Butanediol

SAJ first grade, ≥98.0%

Synonym(s):

1,3-Butylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2CH2OH
CAS Number:
Molecular Weight:
90.12
Beilstein:
1731276
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

grade

SAJ first grade

vapor density

3.1 (20 °C, vs air)

vapor pressure

0.06 mmHg ( 20 °C)

Assay

≥98.0%

form

liquid

autoignition temp.

741 °F

availability

available only in Japan

refractive index

n20/D 1.44 (lit.)

pH

6.0-7.0 (20 °C)

bp

203-204 °C (lit.)

density

1.005 g/mL at 25 °C (lit.)

SMILES string

CC(O)CCO

InChI

1S/C4H10O2/c1-4(6)2-3-5/h4-6H,2-3H2,1H3

InChI key

PUPZLCDOIYMWBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mário T S Rosado et al.
The journal of physical chemistry. A, 113(26), 7499-7507 (2009-04-25)
The complete conformational space of monomeric 1,3-butanediol has been characterized theoretically, and 73 unique stable conformers were found at the MP2/6-311++G(d,p) level. These were classified into nine families whose members share the same heavy atom backbone configurations and differ in
Tamara Kujawska et al.
Ultrasonics, 51(8), 997-1005 (2011-07-05)
This work addresses the difficulties in the measurements of the nonlinear medium parameter B/A and presents a modification of the finite amplitude method (FAM), one of the accepted procedures to determine this parameter. The modification is based on iterative, hybrid
Su Jeong Kim et al.
Nucleosides, nucleotides & nucleic acids, 22(11), 2003-2011 (2003-12-19)
Two optically active phosphoramidite monomers for modified oligodeoxyribonucleotides were prepared. These monomers were then introduced into dodecanucleotides in the middle of the sequences. The modified dodecanucleotides were characterized by various analytical methods including MALDI-TOF mass spectrometry and Tm values were
P A Pugh et al.
Animal reproduction science, 58(1-2), 9-22 (2000-03-04)
This study examined the effects of adding a macromolecule, polyvinylpyrrolidone (10% PVP) and a sugar (0.3 M trehalose) to vitrification solutions (VS) containing either one (40% ethylene glycol [EG], two (25% EG+25% DMSO) or three (20% EG+20% DMSO+10% 1, 3-butanediol
Biagio Velino et al.
The journal of physical chemistry. A, 115(34), 9585-9589 (2011-03-23)
The rotational spectra of five conformers of 1,3-butandiol have been measured by pulsed jet Fourier transform microwave spectroscopy. All of them are stabilized by an internal hydrogen bond and all of them have a GG' or a G'G arrangement of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service