Skip to Content
Merck
All Photos(1)

Key Documents

03-3360

Sigma-Aldrich

Bromine

JIS special grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Br2
CAS Number:
Molecular Weight:
159.81
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

grade

JIS special grade

vapor density

7.14 (vs air)

vapor pressure

175 mmHg ( 20 °C)
671 mmHg ( 55 °C)

Assay

≥99.0%

form

liquid

availability

available only in Japan

resistivity

7.8E18 μΩ-cm, 20°C

bp

58.8 °C (lit.)

mp

−7.2 °C (lit.)

density

3.119 g/mL at 25 °C (lit.)

SMILES string

BrBr

InChI

1S/Br2/c1-2

InChI key

GDTBXPJZTBHREO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teresa M Bisson et al.
Environmental science & technology, 46(21), 12186-12193 (2012-10-02)
Recent laboratory and field-scale experiments demonstrated the potential for brominated industrial solid waste from biomass combustion (Br-Ash) to be an efficient, cost-effective alternative to activated carbon for capturing mercury from coal-fired power plants. To develop this attractive alternative technology to
Bromine-77 and iodine-123 radiopharmaceuticals.
G Stöcklin
The International journal of applied radiation and isotopes, 28(1-2), 131-147 (1977-01-01)
Nickel-butadiene catalytic system for the cross-coupling of bromoalkanoic acids with alkyl Grignard reagents: a practical and versatile method for preparing fatty acids.
Takanori Iwasaki et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(9), 2956-2960 (2013-02-02)
Kimberly T Barrett et al.
Journal of the American Chemical Society, 135(8), 2963-2966 (2013-02-16)
We report the enantioselective synthesis of atropisomeric benzamides employing catalytic electrophilic aromatic substitution reactions involving bromination. The catalyst is a simple tetrapeptide bearing a tertiary amine that may function as a Brønsted base. A series of tri- and dibrominations were
Vitalij V Levin et al.
Organic letters, 15(4), 917-919 (2013-02-02)
Reactions of difluorocarbene with benzyl and alkylzinc halides leading to fluorinated organozinc species have been described. The generated α-difluorinated organozinc reagents are reasonably stable in solution and can be quenched with external electrophiles (iodine, bromine, proton), affording compounds containing the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service