Skip to Content
Merck
All Photos(1)

Key Documents

PX1535

Supelco

Potassium Oxalate, Monohydrate

Synonym(s):

Potassium Oxalate

Sign Into View Organizational & Contract Pricing


About This Item

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

Assay

98.5-101.0%

form

crystals

impurities

≤0.001% Iron (Fe)
≤0.002% Ammonium (NH4)
≤0.002% Chloride (Cl)
≤0.002% Heavy metals (as Pb)
≤0.01% Insoluble matter
≤0.01% Sulfate (SO4)
≤0.02% Sodium (Na)

color

white

pH

7.0-8.5 (20 °C, 50 g/L in H2O)

mp

>138 °C (decomposes before melting)

solubility

392 g/L (anhydrous substance)

storage temp.

room temp

Application

Meets ACS Specifications GR ACS

Analysis Note

Ammonium (NH4):0.002% max
Assay:98.5-101.0%
Chloride (Cl):0.002% max
Color:White
Form:Crystals
Heavy metals (as Pb):0.002% max
Insoluble matter:0.01% max
Iron (Fe):0.001% max
Neutrality:To pass test
Sodium (Na):0.02% max
Substances darkened by hot sulfuric acid:To pass test
Sulfate (SO4):0.01% max

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service