Skip to Content
Merck
All Photos(1)

Key Documents

PC130

Sigma-Aldrich

Anti-Rad51 (Ab-1) Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-Rad51, Rabbit Anti-Rad51, Rad51 Detection Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

mouse, rat, human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... RAD51(5888)

General description

Affinity purified rabbit polyclonal antibody. Recognizes the ~43 kDa Rad51 protein.
Recognizes the ~43 kDa Rad51 protein in ovarian tissue.
The bacterial RecA and the yeast RAD51 proteins are involved in homologous recombination and DNA repair. The human RAD51 protein is the homologue of these two proteins. While the modular structure of these proteins is conserved and the homology is very strong with the bacterial protein there are functional differences seen with RAD51 including an ATP hydrolysis rate 1/10 that seen with RecA. Although RAD51 is a nuclear protein and is largely absent from the cytoplasm and nucleoli, the protein lacks a nuclear localization signal and requires other proteins in a complex to be translocated into the nucleus. Treatment of cells with DNA synthesis inducing agents will cause an increase in the levels of RAD51 protein which then undergo a shift in distribution becoming focal in nature. This focal distribution may be blocked by inhibitors of transcription. RAD51 can be shown to interact directly with the chromosomes and during the process of meiosis, its distribution of RAD51 undergoes dynamic change. RAD51 has been shown to interact with a wide variety of different proteins that include the wild-type form of p53, BRCA2, and RAD52, which specifically modulates its activity.
This Anti-Rad51 (Ab-1) Rabbit pAb is validated for use in Immunoblotting, Immunoprecipitation, Immunocytochemistry for the detection of Rad51 (Ab-1).

Immunogen

Human
recombinant, human Rad51

Application




Immunoblotting (1:2500)
Immunoprecipitation (40 l/500g total protein from lysate)
Immunocytochemistry (1:500)

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 100 mM Tris-Glycine, pH 7.4

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
A431 cells, HeLa cells

Other Notes

Brendel, V., et al. 1997. J. Mol. Evol.44, 528.
Boulikas, T., et al. 1997. Anticancer Res.17, 843.
Gupta, R.C., et al. 1997. Proc. Natl. Acad. Sci. USA94, 463.
Mizuta, R., et al. 1997. Proc. Natl. Acad. Sci. USA 94, 6927.
Sharan, S.K., et al. 1997. Nature386, 304.
Li, M.J., et al. 1996. Proc. Natl. Acad. Sci. USA93, 10222.
Shen, Z., et al. 1996. J. Biol. Chem.271, 148.
Sturzbecher, H.W., et al. 1996. EMBO J. 15, 1992.
Tashiro, S., et al. 1996. Oncogene12, 2165.
Ashley, T., et al. 1995. Chromosoma104, 19.
Haaf, T., et al. 1995. Proc. Natl. Acad. Sci. USA92, 2298.
Due to a high degree of sequence identity, this antibody is also expected to cross-react with bovine, equine, and canine RAD51. Antibody should be titrated for individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nuria Coll-Bonfill et al.
Proteomics, 20(5-6), e1800406-e1800406 (2019-12-14)
Hutchinson Gilford progeria syndrome (HGPS) is a devastating accelerated aging disease caused by LMNA gene mutation. The truncated lamin A protein produced "progerin" has a dominant toxic effect in cells, causing disruption of nuclear architecture and chromatin structure, genomic instability
Roberto de la Fuente et al.
PLoS genetics, 3(11), e198-e198 (2007-11-07)
In most eutherian mammals, sex chromosomes synapse and recombine during male meiosis in a small region called pseudoautosomal region. However in some species sex chromosomes do not synapse, and how these chromosomes manage to ensure their proper segregation is under
Karen J Ouyang et al.
PLoS biology, 7(12), e1000252-e1000252 (2009-12-04)
The gene mutated in Bloom's syndrome, BLM, is important in the repair of damaged replication forks, and it has both pro- and anti-recombinogenic roles in homologous recombination (HR). At damaged forks, BLM interacts with RAD51 recombinase, the essential enzyme in
Yue Sun et al.
Cell death & disease, 11(4), 292-292 (2020-04-29)
Genome instability is the fundamental hallmark of malignant tumors. Tumor suppressors often play a role in maintaining genome stability. Our previous genetic screen identified inositol polyphosphate 4-phosphatase type B (INPP4B), primarily hydrolyzing phosphatidylinositol 3, 4-disphosphate, is a potential tumor suppressor
Céline Jacquemont et al.
Cancer research, 67(15), 7395-7405 (2007-08-03)
Proteasome inhibitors sensitize tumor cells to DNA-damaging agents, including ionizing radiation (IR), and DNA cross-linking agents (melphalan and cisplatin) through unknown mechanisms. The Fanconi anemia pathway is a DNA damage-activated signaling pathway, which regulates cellular resistance to DNA cross-linking agents.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service