Skip to Content
Merck
All Photos(1)

Key Documents

MABX5542

Sigma-Aldrich

Anti-Human hemoglobin (10mg) KC

from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human

technique(s)

ELISA: suitable
immunodiffusion: suitable

isotype

IgG1

General description

Hemoglobin (Hb) is oxygen-binding metalloprotein that is present in the red blood cells. The most abundant Hb is HbA tetramer that is made up of two α, two β chains and the nonprotein heme group. Antibodies againsy Hemoglobulin are routinely used in diagnostics to detect blood in fecal matter (Fecal Occult Blood Test) which is a commonly used test to screen colorectal cancer.

Specificity

Specific for Human hemoglobin. Negative for hemoglobin from pig, bovine, goat, sheep, horse, chicken, rabbit, etc

Immunogen

corresponding to Human Human hemoglobin.

Application

Detect Human hemoglobin using this mouse monoclonal Anti-Human hemoglobin (10mg) KC Antibody, Cat. No. MABX5542-10KC, validated for use in ELISA and Lateral Flow Assay.
Research Category
Apoptosis & Cancer
Suggested antibody pairs MABX5541-1KC and MABX5542-1KC

Quality

Tested

Physical form

Format: Purified
Ion-exchange chromatography
Purified mouse monoclonal in buffer containing 50 mM NaCl, 10 mM PBS, pH 8.0, 0.1 % sodium azide (NaN₃).

Storage and Stability

2-8°C. For long-term storage, freeze at -20°C.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service