Skip to Content
Merck
All Photos(1)

Key Documents

MABT108

Sigma-Aldrich

Anti-TNF-alpha induced protein 6 (TSG-6) Antibody, clone NG3

clone NG3, from mouse

Synonym(s):

Tumor necrosis factor-inducible gene 6 protein, TNF-stimulated gene 6 protein, TSG-6, Tumor necrosis factor alpha-induced protein 6, TNF alpha-induced protein 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

NG3, monoclonal

species reactivity

human, mouse

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

Tumor Necrosis Factor-alpha (TNF-alpha), also known as TSG-6, is a potent lymphoid factor that exerts cytotoxic effects on a wide range of tumor cells and certain other target cells. The mature form of human TNF-alpha has 157 amino acid residues. TNF-alpha induced protein 6 may be involved in cell-cell and cell-matrix interactions during normal inflammatory responses and during tumorigenesis.

Specificity

Published to react with human TSG-6 (Nagyeri, G., et al. (2011). JBC. PMID: 21566135).

Immunogen

Epitope: Unknown
GST-tagged recombinant protein corresponding to mouse TSG-6.

Application

Detect TNF-alpha induced protein 6 (TSG-6) using this Anti-TNF alpha Antibody, clone NG3 validated for use in WB, ELISA. This TNF alpha antibody has been tested in Human and Mouse.
ELISA: A previous lot of this antibody was used in capture ELISA to demonstrate a typical standard curve for measuring TSG6 from normal (naïve) serum.

Western Blot Analysis: A previous lot of this antibody was show to detect human or mouse samples (Nagyeri, G., et al. (2011). JBC. PMID: 21566135).
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Quality

Evaluated by Western Blot in mouse ovary tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected TSG-6 in 10 µg of mouse ovary tissue lysate.

Target description

~29 kDa observed.
An uncharacterized band appears at ~50 kDa in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse ovary tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fiona E Watt et al.
Arthritis & rheumatology (Hoboken, N.J.), 68(9), 2129-2140 (2016-03-19)
To investigate whether molecules found to be up-regulated within hours of surgical joint destabilization in the mouse are also elevated in the analogous human setting of acute knee injury, how this molecular response varies between individuals, and whether it is
F E Watt et al.
Osteoarthritis and cartilage, 28(3), 324-333 (2020-01-07)
Surgical knee joint distraction (KJD) leads to clinical improvement in knee osteoarthritis (OA) and also apparent cartilage regeneration by magnetic resonance imaging. We investigated if alteration of the joint's mechanical environment during the 6 week period of KJD was associated
Chiara Di Santo et al.
International journal of molecular sciences, 24(6) (2023-03-30)
The identification of novel targets to modulate the immune response triggered by cerebral ischemia is crucial to promote the development of effective stroke therapeutics. Since tumour necrosis factor (TNF)-α-stimulated gene 6 (TSG-6), a hyaluronate (HA)-binding protein, is involved in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service