Skip to Content
Merck
All Photos(3)

Key Documents

MAB5206

Sigma-Aldrich

Anti-Amyloid Antibody, β 1-16, clone DE2

culture supernatant, clone DE2, Chemicon®

Synonym(s):

Anti-AAA, Anti-ABETA, Anti-ABPP, Anti-AD1, Anti-APPI, Anti-CTFgamma, Anti-CVAP, Anti-PN-II, Anti-PN2, Anti-alpha-sAPP, Anti-preA4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

DE2, monoclonal

species reactivity

monkey, bovine, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... APP(351)

Application

Anti-Amyloid Antibody, β 1-16, clone DE2 is an antibody against Amyloid for use in ELISA, WB, IH(P).
Western blot: 1:10-1:300 Immunohistochemistry: 1:10-1:200 on formaldehyde fixed, paraffin embedded tissue, requires formic acid or microwave pretreatment to stain Alzheimer′s brain senile plaques. ELISA Immunoprecipitation

Optimal working dilutions must be determined by end user.

Physical form

Culture supernatant. Liquid containing 0.02% sodium azide.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander J Ehrenberg et al.
Acta neuropathologica, 141(5), 651-666 (2021-03-08)
The farnesyltransferase inhibitor, Lonafarnib, reduces tau inclusions and associated atrophy in familial tauopathy models through activation of autophagy, mediated by the inhibition of farnesylation of the Ras GTPase, Rhes. While hinting at a role of Rhes in tau aggregation, it
Mélanie Sipion et al.
Scientific reports, 13(1), 19828-19828 (2023-11-15)
Photobiomodulation (PBM), the process of exposing tissue to red or near-infrared light, has become a topic of great interest as a therapy for diverse pathologies, including neurodegenerative disorders. Here, we aimed to evaluate the potential beneficial effect of PBM on
S100A9 knockout decreases the memory impairment and neuropathology in crossbreed mice of Tg2576 and S100A9 knockout mice model.
Kim, HJ; Chang, KA; Ha, TY; Kim, J; Ha, S; Shin, KY; Moon, C; Nacken, W; Kim, HS; Suh, YH
Testing null
Avindra Nath et al.
Neuromolecular medicine, 3(1), 29-39 (2003-04-01)
Studies of amyloid precursor protein transgenic mice suggest that immune responses to amyloid beta peptide (Abeta) may be instrumental in the removal of plaques from the brain, but the initial clinical trial of an Abeta vaccine in patients with Alzheimer
Tomàs Jordà-Siquier et al.
Alzheimer's & dementia : the journal of the Alzheimer's Association, 18(11), 2099-2116 (2022-01-26)
In Alzheimer's disease (AD), the distribution of the amyloid precursor protein (APP) and its fragments other than amyloid beta, has not been fully characterized. Here, we investigate the distribution of APP and its fragments in human AD brain samples and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service