Skip to Content
Merck
All Photos(2)

Key Documents

MAB4419A4

Sigma-Aldrich

Anti-OCT-4 [POU5F1] Antibody, clone 7F9.2, Alexa Fluor 488 conjugate

clone 7F9.2, from mouse, ALEXA FLUOR 488

Synonym(s):

POU domain class 5, transcription factor 1, Octamer-binding protein 3, Oct-3, Octamer-binding protein 4, Oct-4, Octamer-binding transcription factor 3, OTF-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified antibody

antibody product type

primary antibodies

clone

7F9.2, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... POU5F1(5460)
mouse ... Pou5F1(18999)

General description

Octamer-4 (Oct-4), a member of the POU family of transcription factors, has been demonstrated to be vital for the formation of self-renewing pluripotent stem cells. During embryogenesis, expression of Oct-4 is limited to pluripotent cells of the inner cell mass (ICM) that contribute to the formation of all fetal cell types. This relationship between Oct-4 and pluripotency has seen this transcription factor emerge as a marker of pluripotent stem cells. Undifferentiated human and murine pluripotent Embryonic Stem (ES) and Embryonic Carcinoma (EC) cells express Oct-4. Additionally, murine Embryonic Germ (EG) cells are also known to express Oct-4. Following stem cell differentiation, the level of Oct-4 expression decreases. Oct-4 has been identified as one of the main transcription factors required to reprogram somatic cells into induced pluripotent stem cells (iPS cells).

Application

Anti-OCT-4 [POU5F1] Antibody, clone 7F9.2, Alexa Fluor 488 conjugate is an antibody against OCT-4 [POU5F1] for use in ICC.

Quality

Evaluated by Immunocytochemistry in mouse embryonic stem cells (SCR012). Immunocytochemsitry Analysis: A 1:100 dilution of this antibody detected Oct-4 in mouse embryonic stem cells (SCR012).

Target description

The uncojugated parent antibody (Catalog No. MAB4419) has an observed MW at 39 kDa

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Li et al.
PloS one, 15(6), e0234262-e0234262 (2020-06-10)
p53 is one of the most extensively studied proteins in cancer research. Mutations in p53 generally abolish normal p53 function, and some mutants can gain new oncogenic functions. However, the mechanisms underlying p53 mutation-driven cancer remains to be elucidated. Our
Brian C Del Rosario et al.
Developmental cell, 43(3), 359-371 (2017-11-07)
X-chromosome inactivation (XCI) silences one X chromosome in the female mammal and is essential to peri-implantation development. XCI is thought to be cell autonomous, with all factors required being produced within each cell. Nevertheless, external cues may exist. Here, we
Wanqiao Wang et al.
British journal of haematology, 202(2), 328-343 (2023-05-05)
Juvenile myelomonocytic leukaemia (JMML) is an aggressive paediatric leukaemia characterized by mutations in five canonical RAS pathway genes, including the NF1 gene. JMML is driven by germline NF1 gene mutations, with additional somatic aberrations resulting in the NF1 biallelic inactivation
Tingting Cheng et al.
Oncotarget, 8(5), 7814-7826 (2016-12-22)
Primordial germ cells (PGCs) derived from human embryonic stem cells (hESCs) represent as a desirable experimental model as well as a potential strategy for treating male infertility. Here, we developed a simple and feasible method for differentiation of PGCs from
Melissa Conti Mazza et al.
Stem cell research, 55, 102506-102506 (2021-08-23)
Mutations in the oncogene PARK7, which codes for DJ-1, have been associated with early-onset autosomal recessive Parkinson's disease (PD); however, the exact role of DJ-1 in PD remains elusive. Fibroblasts from a PD patient with a uniparental disomy, 1 bp deletion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service