Skip to Content
Merck
All Photos(1)

Key Documents

HX0298

Supelco

Hexane, mixture of isomers

≥98.5% (hexane isomers + methylcyclopentane), suitable for gas chromatography (GC), Non-UV OmniSolv®

Synonym(s):

Hexanes, Hexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14
CAS Number:
Molecular Weight:
86.18
MDL number:
UNSPSC Code:
12191502
EC Index Number:
203-777-6

product name

Hexanes, 98.5%,, for GC analyses, Non-UV OmniSolv®

vapor pressure

160 hPa ( 20 °C)

Quality Level

product line

OmniSolv®

Assay

≥98.5% (hexane isomers + methylcyclopentane)
≥99.8% (total aliphatic hydrocarbons less than C8)

form

liquid

technique(s)

gas chromatography (GC): suitable

impurities

≤0.01% Water (H2O)
≤2 ng/L Capillary ECD responsive substances (as PCNB)
≤3 μg/L Capillary FID responsive substances (as decane)

evapn. residue

≤1 ppm

color

APHA: ≤10

bp

69 °C/1013 hPa

mp

-94.3 °C

transition temp

flash point -22 °C

solubility

0.0095 g/L

density

0.66 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

for GC analyses, Non-UV OmniSolv®

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (n-hexane)Report%Assay (total aliphatic hydrocarbons less than C8)99.8% minAssay (total hexane isomers + methylcyclopentane)98.5% minBoiling range66.1-69.4°CCapillary ECD responsive substances (as PCNB)2ng/L maxCapillary FID responsive substances (as decane)3µg/L maxColor (APHA)10 maxEvaporation residue1ppm maxFiltered through 0.2 µm filterTo pass testFormClear liquidInfrared SpectrumConformsRefractive index (at 20°C)Water (H2O)0.01% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service