Skip to Content
Merck
All Photos(1)

Key Documents

AP183F

Sigma-Aldrich

Goat Anti-Rat IgG Antibody, FITC conjugate, Species Adsorbed

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

FITC conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

Application

Research Category
Secondary & Control Antibodies
Research Sub Category
Secondary Antibodies Adsorbed for Dual Labeling
This Goat anti-Rat IgG Antibody, FITC conjugate, Species Adsorbed is validated for use in IF for the detection of Rat IgG.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Ishikawa et al.
Communications biology, 4(1), 409-409 (2021-04-09)
Organogenesis and regeneration are fundamental for developmental progress and are associated with morphogenesis, size control and functional properties for whole-body homeostasis. The liver plays an essential role in maintaining homeostasis of the entire body through various functions, including metabolic functions
Xing-Qi Zhao et al.
Frontiers in physiology, 13, 808272-808272 (2022-08-13)
Background: Previous studies had reported that vitamin D receptor (VDR) gene polymorphisms were related to the development of several inflammatory disorders. However, potential links between such variations and the risk of developing a bone infection and underlying mechanisms remain unclear.
Laura Conti et al.
Contrast media & molecular imaging, 8(4), 350-360 (2013-04-25)
Despite spontaneous tumor growth in genetically engineered mice being one of the most recognized tools for the in vivo evaluation of novel diagnostic and therapeutic anticancer compounds, monitoring early stage lesions in live animals is a goal that has yet
Aakash Koppula et al.
Cancers, 14(2) (2022-01-22)
Circular RNAs (circRNAs) are regulatory RNAs which have recently been shown to have clinical significance in several diseases, including, but not limited to, various cancers, neurological diseases and cardiovascular diseases. The function of such regulatory RNAs is largely dependent on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service