Skip to Content
Merck
All Photos(1)

Key Documents

AB3209

Sigma-Aldrich

Anti-Oct-4 Antibody

Chemicon®, from rabbit

Synonym(s):

POU5F1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

input

sample type induced pluripotent stem cell(s)
sample type: human embryonic stem cell(s)
sample type: mouse embryonic stem cell(s)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... POU5F1(5460)

General description

Expression of the POU-domain transcription factor Octamer-4 (Oct-4) is widely regarded as a hallmark of pluripotent stem cells. The relationship of Oct-4 to pluripotent stem cells is indicated by its tightly restricted expression to undifferentiated pluripotent stem cells. Upon differentiation to somatic lineages, the expression of Oct-4 disappears rapidly. Unlike the majority of pluripotent stem cell markers, the biological role of Oct-4 has been well characterized. Studies performed in mice point to the critical role of Oct-4 in the establishment and/or maintenance of pluripotent stem cells in an uncommitted state.

Specificity

Recognizes Oct4 (POU5F1). The molecular weight of the protein is 38,601 Daltons.

Immunogen

Synthetic peptide, amino acids 282-297of Oct4.

Application

Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation
This Anti-Oct-4 Antibody is validated for use in IC, IH, WB for the detection of 41186.
Western blot: 1:200-1:1,000 using ECL. Oct-4 in cells typically shows ~40-44kDa band or doublet.

Immunohistochemistry: 1:200-1:1,000.

Immunocytochemistry: 1:200-1:1,000.

Optimal working dilutions must be determined by the end user.

Target description

44 kDa

Physical form

Affinity Purified immunoglobulin. Precipitated antibody in a solution of 50% saturated ammonium sulfate and PBS containing no preservatives.
ImmunoAffinity Purified

Storage and Stability

Maintain unopened vial at -70°C for up to 6 months. Avoid repeated freeze/thaw cycles.

The rehydrated antibody solutions can be stored undiluted at 2–8°C for 2 months without any significant loss of activity. Note, the solution is not sterile, thus care should be taken if product is stored at 2–8°C. For storage at -20°C, the addition of an equal volume of glycerol can be used, however, it is recommended that ACS grade or higher glycerol be used, as significant loss of activity can occur if the glycerol used is not of high quality.

For freezing, it is recommended that the rehydrated antibody solution be further diluted 1:1 with a 2% BSA (fraction V, highest-grade available) solution made with the rehydration buffer. The resulting 1% BSA/antibody solution can be aliquoted and stored frozen at -70°C for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Pluripotent human embryonic stem (ES) cells

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chawalit Siriboon et al.
PloS one, 10(2), e0118165-e0118165 (2015-02-14)
We attempted to isolate ES cell lines using inner cell masses from high-quality cloned porcine blastocysts. After being seeded onto feeders, embryos had better (P < 0.05) attachment, outgrowth formation and primary colonization in both 2× and 3× aggregated cloned
Difei Dong et al.
Gene expression, 15(4), 153-162 (2012-07-13)
FoxA1 belongs to the fork head/winged-helix transcription factor family and participates in stimulating neuronal differentiation of pluripotent stem cells at early stages. To explore the biological roles of FoxA1 during this process, the stable expression of a GFP-FoxA1 fusion protein
Neuronatin promotes neural lineage in ESCs via Ca(2+) signaling.
Lin, HH; Bell, E; Uwanogho, D; Perfect, LW; Noristani, H; Bates, TJ; Snetkov, V; Price, J; Sun, YM
Stem Cells null
Max Villa et al.
Biomedical microdevices, 12(2), 253-261 (2009-12-17)
We present optimal perfusion conditions for the growth of primary mouse embryonic fibroblasts (mEFs) and mouse embryonic stem cells (mESCs) using a microfluidic perfusion culture system. In an effort to balance nutrient renewal while ensuring the presence of cell secreted
Filipa Vlahova et al.
Scientific reports, 9(1), 8126-8126 (2019-06-01)
Expression of OCT4A is one of the hallmarks of pluripotency, defined as a stem cell's ability to differentiate into all the lineages of the three germ layers. Despite being defined as non-tumorigenic cells with high translational potential, human mid-trimester amniotic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service