Skip to Content
Merck
All Photos(1)

Key Documents

AB15328

Sigma-Aldrich

Anti-Olig2 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... OLIG2(10215)

Specificity

Oligodendrocyte marker Olig2. By Western blot the antibody recognizes a band at ~32 kDa in rat brain extract.

Immunogen

Synthetic peptide from rat.

Application

Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers
This Anti-Olig2 Antibody is validated for use in WB for the detection of Olig2.

Physical form

ImmunoAffinity Purified
Liquid in PBS containing 0.1% sodium azide.

Storage and Stability

Maintain at 2-8degC in undiluted aliquots for up to 6 months after date of receipt.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Blake Chancellor et al.
Acta neuropathologica, 142(2), 295-321 (2021-05-22)
Chronic traumatic encephalopathy (CTE) is a progressive tauopathy found in contact sport athletes, military veterans, and others exposed to repetitive head impacts. White matter rarefaction and axonal loss have been reported in CTE but have not been characterized on a
Embryonic stem cell-based mapping of developmental transcriptional programs.
Mazzoni, EO; Mahony, S; Iacovino, M; Morrison, CA; Mountoufaris, G; Closser, M; Whyte et al.
Nature Methods null
A Comprehensive Profile of ChIP-Seq-Based Olig2 Target Genes in Motor Neuron Progenitor Cells Suggests the Possible Involvement of Olig2 in the Pathogenesis of Amyotrophic Lateral Sclerosis.
Satoh, J; Asahina, N; Kitano, S; Kino, Y
Journal of Central Nervous System Disease null
Ana Bernal-Chico et al.
Glia, 63(1), 163-176 (2014-08-19)
The endocannabinoids 2-araquidonoylglycerol (2-AG) and anandamide (AEA) are bioactive lipids crucially involved in the regulation of brain function in basal and pathological conditions. Blockade of endocannabinoid metabolism has emerged as a promising therapeutic strategy for inflammatory diseases of the central
Xiaoyang Wang et al.
Journal of child neurology, 24(9), 1171-1178 (2009-09-12)
Preterm infants exposed to inflammation are at increased risk of white matter injury and/or cerebral palsy. To investigate the effect of chronic inflammation on the developing white matter, we administered low-dose lipopolysaccharide once a day from postnatal days 3 to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service