Skip to Content
Merck
All Photos(1)

Key Documents

8.41249

Sigma-Aldrich

Tetramethylammonium acetate

(50% solution in water) for synthesis

Synonym(s):

Tetramethyl ammonium acetate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352107
NACRES:
NA.22

Quality Level

form

liquid

density

1.000 g/cm3 at 20 °C

storage temp.

2-30°C

Related Categories

Analysis Note

Assay (HClO₄): 45.0 - 55.0 %

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service